Sodium metasilicate, nonahydrate 44 - 47.5% total solids (500g) (AC419595000)

YO-88356-78
Loading price...

Usually ships in 14 days.
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 13517-24-3
    Formula weight 284.19
    Empirical formula Na2SiO3.9H2O
    Linear formula Na2SiO3.9H2O
    Hazard symbol Corrosive
    Manufacturer number AC419595000
    Quantity 500g





    Technical Resources
     
    Related Products for Sodium metasilicate, nonahydrate 44 - 47.5% total solids (500g) (AC419595000)
    Image
      Product Detail
    SKU

    Sodium Sulfide Nonahydrate (Cryst./Certified) (2.5kg) (S425212)

    Category: Sodiump to Sodiumz
    Description: Sodium Sulfide Nonahydrate
    Brand: Cole-Parmer


    Carousel stand for pipettors (22 44 490-5)

    Category: Eppendorf Titermate Multichannel Pipettors
    Description: Carousel stand for pipettors
    Brand: Eppendorf


    Sodium Hydroxide Solution (0.2N (N/5)/Certified) (1 liter)

    Category: Sodiumh to Sodiumo
    Description: Sodium Hydroxide Solution
    Brand: Hydroxide


    Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals) (500g)

    Category: Sodiump to Sodiumz
    Description: Sodium Phosphate Dibasic Heptahydrate
    Brand: Cole-Parmer


    2-Amino-5-chloropyridine 98% (500g) (AC103415000)

    Category: Aminoc
    Description: 2-Amino-5-chloropyridine
    Brand: Cole-Parmer


    3-Methyl-1-phenyl-2-pyrazolin-5-one 99% (500g) (AC127465000)

    Category: Methylp to Methylq
    Description: 3-Methyl-1-phenyl-2-pyrazolin-5-one
    Brand: Cole-Parmer


    Sodium Metasilicate Nonah 25g (AC419590250)

    Category: Chemicals, Reagents, & Lubricants
    Description: Specialty Chemicals
    Brand: Cole-Parmer


    Sodium Sulfide Nonahydrate 25g (YO-88363-05)

    Category: Chemicals, Reagents, & Lubricants
    Description: Specialty Chemicals
    Brand: Cole-Parmer


    Poly(acrylamide), granular (non-ionic), average M.W. 5 to 6.000.000 (500g)

    Category: Platina to Potash
    Description: Poly(acrylamide)
    Brand: Cole-Parmer


    Solid Color-Coded Silicone Stoppers, Standard Size 10.5, Blue; 5/Bag

    Category: Stoppers
    Description: Solid Color-Coded Stoppers, Silicone
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Sodium metasilicate, nonahydrate 44 - 47.5% total solids (500g) (AC419595000)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Sodium Metasilicate Nonah 25g (AC419590250)  |  Next: Sodium metatungstate hydrate, crystalline (250g) (YO-88279-58)