Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermocouple Temperature Controller Type K and J 110V C
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-93520-02
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17090-18
NIST-Traceable Calibration; Panel Meters
$139.61 AUD/EACH/EACH
Usually ships in 24 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Controllers-Panel Mount
Share
|
Thermocouple Temperature Controller, Type K and J, 110V, °C (TCS-4011)
YO-93520-02
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Digital display and adjustable deadband for a very economical price
Powerful 16 A, SPDT relay can run up to a 3/4-hp compressor
Feature password protection and error messages
These controllers are ideal for on/off control applications where a digital display and an adjustable deadband are needed over a limited temperature range. Use with packaging equipment, food cooling and chilling equipment, and more. Each controller has three push buttons for programming parameters including set point, deadband, cycle time, and ambient probe adjustment. Programmable password protection prevents accidental changes to settings. Multiple units can be set with the same program with the use of a “configuration key” (sold separately). The bright 3-digit LED display is easy to read even in dim areas Controllers are available with a thermocouple or thermistor input; the thermocouple version features two selectable alarm conditions and internal buzzer. The FM-approved limit models include field-selectable automatic or manual reset, as well as front-panel or external input reset option. Order probes separately.
What's Included:
temperature controller.
Specifications
Product Type
Temperature control
Control input (thermocouple)
Yes
Control output 1
Relay: SPDT, 16 A at 250 VAC
Alarm output
Yes
Min temperature (º C)
-10
Max temperature (º C)
55
Min temperature (º F)
14
Max temperature (º F)
131
Dimensions (" W)
2.99
Dimensions (" H)
1.34
Dimensions (" D)
2.35
Display
3-digit LED, red, 1/2" H
Panel cutout
2.8" W x 1.14" H (71 x 29 mm)
Temp accuracy
±1°F (±1°C)
Resolution
1°F (1°C)
CE Compliance
Yes
Enclosure rating
NEMA 4 (IP65)
Brand
Love
Model
TCS-4011
Manufacturer number
TCS-4011
Power (VAC)
110
Accessories
Qty
/
Price
Availability
Configuration Key for Love Controllers
YO-93520-17
EACH
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Panel Meters
YO-17090-18
EACH
Usually ships in 24 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Temperature Control: An Overview and Product Selection Guide
Temperature Controller Features
Electron zap turns flimsy plastic into sturdy shrink wrap
Show All...
Basics of Cartridge Filtration
Bitter Brew
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Related Searches for Thermocouple Temperature Controller, Type K and J, 110V, °C (TCS-4011)
Type J Thermocouple Temperature Controller
Thermocouple Temperature Controller
Micro Panel Mount Thermocouple Temperature Controller
Portable Thermocouple Temperature Controller
Thermocouple Temperature Controller Log
Honeywell Thermocouple Temperature Controller
Temperature Controller Type 5341 3502
Oakton Temperature Controllers Type
Temperature Controller Type T
Temperature Controller 110v
Related Products for Thermocouple Temperature Controller, Type K and J, 110V, °C (TCS-4011)
Image
Product Detail
SKU
Thermocouple Temperature Controller, Type K and J, 110V, °F (TCS-4010)
Category: Controllers-Panel Mount
Description: Love On/Off and Limit Switch Temperature Controllers
Brand:
Love
Model Number:
TCS-4010
YO-93520-01
More Details
Thermocouple Temperature Controller, Type K and J, 230V, °F (TCS-4020)
Category: Controllers-Panel Mount
Description: Love On/Off and Limit Switch Temperature Controllers
Brand:
Love
Model Number:
TCS-4020
YO-93520-06
More Details
Thermocouple Temperature Controller, Type K and J, 230V, °C (TCS-4021)
Category: Controllers-Panel Mount
Description: Love On/Off and Limit Switch Temperature Controllers
Brand:
Love
Model Number:
TCS-4021
YO-93520-07
More Details
Autotuning Benchtop Temperature Controllers; type J; 100-240V
Category: Controllers-Benchtop
Description: Love Autotuning Benchtop Thermocouple Controllers
Brand:
Love
Model Number:
25013-986/J
YO-02110-82
More Details
Digi-Sense 1-Zone benchtop dual display temperature control, type J in (YO-36225-61)
Category: Controllers-Benchtop
Description: Digi-Sense Benchtop PID Temperature Controllers
Brand:
Digi-Sense
YO-36225-61
More Details
Digi-Sense 1-Zone benchtop dual display temperature control, type K in (YO-36225-62)
Category: Controllers-Benchtop
Description: Digi-Sense Benchtop PID Temperature Controllers
Brand:
Digi-Sense
YO-36225-62
More Details
Digi-Sense 1-Zone benchtop dual display temperature control, type T in (YO-36225-63)
Category: Controllers-Benchtop
Description: Digi-Sense Benchtop PID Temperature Controllers
Brand:
Digi-Sense
YO-36225-63
More Details
Digi-Sense 1-Zone benchtop dual display temperature control, type RTD (YO-36225-64)
Category: Controllers-Benchtop
Description: Digi-Sense Benchtop PID Temperature Controllers
Brand:
Digi-Sense
YO-36225-64
More Details
Digi-Sense 2-Zone benchtop dual display temperature control, type J in (YO-36225-71)
Category: Controllers-Benchtop
Description: Digi-Sense Benchtop Twin Temperature Controllers
Brand:
Digi-Sense
YO-36225-71
More Details
Digi-Sense 2-Zone benchtop dual display temperature control, type K in (YO-36225-72)
Category: Controllers-Benchtop
Description: Digi-Sense Benchtop Twin Temperature Controllers
Brand:
Digi-Sense
YO-36225-72
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermocouple Temperature Controller, Type K and J, 110V, °C (TCS-4011)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermocouple Switch/transmitter, 1 To 1000 Mtorr
|
Next: Thermocouple Temperature Controller, Type K and J, 110V, °F (TCS-4010)
31125329-p-content_2 31125329pcontent2