Industrial Thermocouple Element, 6"L, 20 Gauge, K type (5504A-K-20-6-0)

YO-93605-42
Loading price...

Call for availability.
Qty

  • Heavy-duty construction for high temperature use
Elements with in an industrial thermocouple that includes a thermowells that deteriorate over time can be replaced as a more cost effective solution. Each replacement element is made of 20 gauge wire and is encased with ceramic insulators for a greater degree of protection.
Product Type Thermocouple Probe
Min temperature (º F) -200
Max temperature (º F) 2300
Max temperature (º C) 1260
Probe length (in.) 6
Temp range -200 to 2300°F (-129 to1260°C)
Probe type K
Min temperature (º C) -129
Gauge 20 Gauge
Junction grounded
Manufacturer number 5504A-K-20-6-0
Model 5504A-K-20-6-0

Calibrations
Qty/Price
Availability

EACH
Usually ships in 25 days.




Technical Resources
 
Related Products for Industrial Thermocouple Element, 6"L, 20 Gauge, K type (5504A-K-20-6-0)
Image
  Product Detail
SKU

Industrial Thermocouple Element, 6"L, 20 Gauge, J type (5504A-J-20-6-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Industrial Thermocouple Element, 4"L, 20 Gauge, K type (5504A-K-20-4-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Industrial Thermocouple Element, 9"L, 20 Gauge, K type (5504A-K-20-9-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Industrial Thermocouple Element, 12"L, 20 Gauge, K type (5504A-K-20-12-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Industrial Thermocouple Element, 4"L, 20 Gauge, J type (5504A-J-20-4-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Industrial Thermocouple Element, 9"L, 20 Gauge, J type (5504A-J-20-9-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Industrial Thermocouple Element, 12"L, 20 Gauge, J type (5504A-J-20-12-0)

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Elements
Brand: Cole-Parmer


Type K Grounded Industrial Thermocouple Probe - 6" Length, 1/2" Connection

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Probes with Thermowell
Brand: Cole-Parmer


Type K Grounded Industrial Thermocouple Probe, 6" Length, 3/4" Connection

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Probes with Thermowell
Brand: Cole-Parmer


Type J Grounded Industrial Thermocouple Probe, 6" Length, 1/2" Connection

Category: Digi-Sense Thermocouple Industrial Probes
Description: Digi-Sense Thermocouple Industrial Probes with Thermowell
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews