Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Folding Skirt Stopper Rubber 10 2 Dia 50 pk
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-62995-81
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Stoppers
Share
|
Folding Skirt Stopper Rubber 10.2 Dia 50/pk
YO-62995-81
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Two-part folding skirt stoppers are designed to keep your contents secure two ways. The lower section is molded to grip the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim. Ideal for securing glass, metal, and plastic containers regardless of type. Upper hollow plug section can be perforated to exact contents without removing the stopperremains watertight even after several perforations.
The rubber stoppers are ideal for chemical transfer and withstand temperatures from -4°F (-20°C) to 212°F (100°C). Chemical-resistent silicone stoppers are recommended for biological applications and withstand temperatures from -76°F (-60°C) to 428°F (220°C)
Specifications
Product Type
Stoppers
Standard size number
10.2 dia
Top OD (mm)
10.7
Color
Natural
Material
Rubber
Bottom OD (mm)
9.8
Qty/pk
50
Manufacturer number
407010-50
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Physical Properties of Plastics
Physical Properties of Glass
Physical Properties of Stainless Steel
Show All...
Show Less...
Related Searches for Folding Skirt Stopper Rubber 10.2 Dia 50/pk
Folding Skirt Stoppers
Cole-Parmer Two-Hole Stoppers
Cole-Parmer Twistit Stoppers
Silicone Rubber Stoppers
Products of Rubber
Rubber Tubing Lab Supplies
Rubber Glass
Indenti-Plug Identi-Plug Plastic Stoppers
Ebbco W80 50 Filter Paper
50 Ml Sterile Glass Centrifuge Tube
Related Products for Folding Skirt Stopper Rubber 10.2 Dia 50/pk
Image
Product Detail
SKU
Folding Skirt Stopper Rubber 12.7 Dia 50/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-82
More Details
Folding Skirt Stopper Rubber 23.7 Dia 10/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-86
More Details
Folding Skirt Stopper Rubber 7.1 Dia 100/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-80
More Details
Folding Skirt Stopper Rubber 14.9 Dia 20/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-83
More Details
Folding Skirt Stopper Rubber 15.9 Dia 20/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-84
More Details
Folding Skirt Stopper Rubber 19.4 Dia 20/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-85
More Details
Folding Skirt Stopper Rubber 30.7 Dia 5/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-87
More Details
Folding Skirt Stopper Silicone 10.2 Dia 50/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-91
More Details
Folding Skirt Stopper Silicone 12.7 Dia 50/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-92
More Details
Folding Skirt Stopper Silicone 23.7 Dia 10/pk
Category: Stoppers
Description: Folding Skirt Stoppers, Rubber or Silicone
Brand:
Lab Products
YO-62995-96
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Folding Skirt Stopper Rubber 10.2 Dia 50/pk
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Folding impeller, 3-3/4" dia (max) x 5/8" bore dia (000566)
|
Next: Folding Skirt Stopper Rubber 12.7 Dia 50/pk
31125329-p-content_2 31125329pcontent2