Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Ratchet Clamp 0 197 25 pack
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-95785-04
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Flow Control Clamps
Share
|
Ratchet Clamp, 0.197", 25/pack (YO-95785-04)
YO-95785-04
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Open or close clamps with the simple push of the thumb latch
Thumb latch allows for easy opening or closing from any angle
Reduce strain on tubing line due to opening or closing
Tubing pinch clamps provide quick and easy on/off flow control of fluids in flexible tubing. Acetal construction.
Specifications
Product Type
Flow Control Clamps
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for Ratchet Clamp, 0.197", 25/pack (YO-95785-04)
Z-Man Corp Clamps
Flow Control Clamps - Plastic Pinchcock
Laboratory Controller Clamp
Flow Control Clamps
Product Type Flow Control Clamps
Flow Control Clamps - Pinchcocks
Supply Corp
Small Band Tubing Clamps
2.5 Hose Fitting Fittings
Clamp Hose Fittings
Related Products for Ratchet Clamp, 0.197", 25/pack (YO-95785-04)
Image
Product Detail
SKU
Ratchet Clamp, 0.118", 25/pack (YO-95785-00)
Category: Flow Control Clamps
Description: Flow Control Clamps - Plastic Pinchcock
Brand:
Z-Man Corp
YO-95785-00
More Details
Ratchet Clamp, 0.125", 25/pack (YO-95785-02)
Category: Flow Control Clamps
Description: Flow Control Clamps - Plastic Pinchcock
Brand:
Z-Man Corp
YO-95785-02
More Details
Ratchet Clamp, 0.250", 25/pack (YO-95785-06)
Category: Flow Control Clamps
Description: Flow Control Clamps - Plastic Pinchcock
Brand:
Z-Man Corp
YO-95785-06
More Details
Male Pipe Adapter, Polypropylene, 10-32 UNF x 1/16", 25 PACK (YO-30621-04)
Category: Straight barbed to male UNF threaded adapters
Description: Natural polypropylene straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-30621-04
More Details
Male Pipe Adapter, Polypropylene, 10-32 UNF x 1/16", 25 PACK (YO-40621-04)
Category: Barbed to threaded adapters
Description: Straight barbed to male UNF threaded adapters
Brand:
Cole-Parmer
YO-40621-04
More Details
Band Clamps, 1", 25/pack (YO-33350-12)
Category: Custom-Fit Hose Fittings
Description: Hose Fittings
Brand:
Cole-Parmer
YO-33350-12
More Details
Band Clamps, 2", 25/pack (YO-33350-14)
Category: Custom-Fit Hose Fittings
Description: Hose Fittings
Brand:
Cole-Parmer
YO-33350-14
More Details
Band Clamps, 4", 25/pack (YO-33350-16)
Category: Custom-Fit Hose Fittings
Description: Hose Fittings
Brand:
Cole-Parmer
YO-33350-16
More Details
Band Clamps, 6", 25/pack (YO-33350-18)
Category: Custom-Fit Hose Fittings
Description: Hose Fittings
Brand:
Cole-Parmer
YO-33350-18
More Details
Ratchet tubing clamp for up to 1/2" tubing, acetal, 25/pack (YO-06833-00)
Category: Flow Control Clamps
Description: Flow Control Clamps - Ratchet Tubing Clamp
Brand:
Thermoplastic
YO-06833-00
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Ratchet Clamp, 0.197", 25/pack (YO-95785-04)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Ratchet Clamp, 0.125", 25/pack (YO-95785-02)
|
Next: Ratchet Clamp, 0.250", 25/pack (YO-95785-06)
31125329-p-content_2 31125329pcontent2