Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Wiss Heavy Duty Replacement Blades 025 5 Pack
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-97105-55
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Wiss Heavy Duty Replacement Blades, .025", 5 Pack (YO-97105-55)
YO-97105-55
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related Searches for Wiss Heavy Duty Replacement Blades, .025", 5 Pack (YO-97105-55)
Heavy Duty Blades
Heavy-Duty Carboys with Shoulder Handle
Heavy Duty Carts
Heavy Duty Utility Carts
Cole-Parmer Heavy-Duty Plastic Service Carts
Cole-Parmer Azlon Heavy-Duty Tweezers
Thermo Scientific Nalgene Heavy-Duty Vacuum Carboys
Cole Heavy-Duty Plastic Service
Cole-Parmer Drum/Barrel Heavy-Duty Mixers
Oakton Heavy-Duty Industrial Penetration Probes
Related Products for Wiss Heavy Duty Replacement Blades, .025", 5 Pack (YO-97105-55)
Image
Product Detail
SKU
Economical heavy duty carboy with spigot, 5 liter
Category: Square/Rectangular Carboys and Jerricans
Description: Cole-Parmer Heavy-Duty Carboys with Shoulder Handle and Spigots, PP
Brand:
Guaranteed In Stock Program
Model Number:
505634-1
YO-62507-05
More Details
Heavy-Duty, Variable Speed Mixer, 120 VAC (YO-04340-10)
Category: Talboys Mixers
Description: Talboys Heavy-Duty, Variable-Speed Mixer
Brand:
Cole-Parmer
YO-04340-10
More Details
Threaded adapter, nylon, 3/4" NPTM x 1" tubing ID, 5/pack (YO-30904-55)
Category: Barbed to threaded adapters
Description: Straight barbed to male NPT threaded adapters
Brand:
Cole-Parmer
YO-30904-55
More Details
Heavy-Duty Bath, HDPE, 7 gal
Category: Trays
Description: Heavy-Duty Baths, HDPE
Brand:
Cole-Parmer
Model Number:
P339-01-2510
YO-06712-10
More Details
Heavy-Duty Bath, HDPE, 15 gal
Category: Trays
Description: Heavy-Duty Baths, HDPE
Brand:
Cole-Parmer
Model Number:
P337-01-2510
YO-06712-30
More Details
Heavy-Duty Smart™ Container, HDPE, 30 gal
Category: Mobile Bins and Tubs
Description: Heavy-Duty Smart Container, HDPE
Brand:
Cole-Parmer
Model Number:
P300W/C
YO-06740-50
More Details
Economical heavy-duty graduated polypropylene carboy with shoulder han (505634-5)
Category: Square/Rectangular Carboys and Jerricans
Description: Cole-Parmer Heavy-Duty Carboys with Shoulder Handle, PP
Brand:
Guaranteed In Stock Program
Model Number:
505634-5
YO-62507-10
More Details
Thermo Scientific Nalgene heavy-duty polypropylene bottle, 5 L
Category: Heavy-Duty Bottles
Description: Thermo Scientific Nalgene Heavy-Duty Polypropylene Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2126-5000
YO-06257-30
More Details
Heavy- Duty Industrial Pressure Transmitter, 0 to 5 psi; 4 to 20 mA
Category: Pressure Transmitters and Pressure Transducers
Description: Heavy-Duty Industrial Pressure Transmitter
Brand:
Ashcroft
Model Number:
A25PSI
YO-68334-04
More Details
Heavy- Duty Industrial Pressure Transmitter, 30" Hg to 0 psi; 1 to 5 V
Category: Pressure Transmitters and Pressure Transducers
Description: Heavy-Duty Industrial Pressure Transmitter
Brand:
Ashcroft
Model Number:
A20#/VAC
YO-68334-30
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Wiss Heavy Duty Replacement Blades, .025", 5 Pack (YO-97105-55)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Wiss 8" Take-Apart Utility Scissors (YO-97105-57)
|
Next: Wiss Quick Change Confort Grip Utility Knife (YO-97105-53)
31125329-p-content_2 31125329pcontent2