Small Kimwipes wipe dispenser

YO-63528-10
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Keep disposable supplies where they're needed
  • Free valuable drawer space for storage

Wipe Dispensers are ready to mount near instruments, workstations, hoods, and benches. Simply peel adhesive tape backing and press on.
Product Type Organizer
Color White
No. of drawers or compartments 1
Dimensions (" W) 4.625
Dimensions (" H) 3
Dimensions (" D) 4.75





Technical Resources
 
Related Products for Small Kimwipes wipe dispenser
Image
  Product Detail
SKU

Large Kimwipes wipe dispenser

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Parafilm


Modular laboratory organizer, small Kimwipes box dispenser

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Cole-Parmer
Model Number: 63535-11


Large Shur-Wipe wipe dispenser

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Parafilm


Modular laboratory organizer, large Kimwipes box dispenser

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Cole-Parmer
Model Number: 63535-12


Modular laboratory organizer, Parafilm dispenser

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Cole-Parmer
Model Number: 63535-10


Modular laboratory organizer, glove box dispenser

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Cole-Parmer
Model Number: 63535-13


Glove box holder for one box

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Parafilm


Glove box holder for two boxes

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Parafilm


Glove box holder for three boxes

Category: Laboratory Organizers
Description: Glove, Kimwipes & Parafilm Dispensers
Brand: Parafilm


Kimwipes EX-L Wipes, 4-1/2" X 8-1/2", 280/box, 60 boxes/case

Category: Clean Room Wipes
Description: Kimberly-Clark Critical Task Wipes
Brand: Kimberly-Clark
Model Number: 3235713 OR 34155


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    can this dispenser be hung on a wall?
    Asked on 5/13/2011 by Anonymous

    1 answer

    • Staff Reviewer

      A:

      Yes, these come with a double sided adhesive and can be hung on a wall.

      Answered on 5/16/2011 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews
Customers Who Bought This Item Also Bought

Small Kimwipes wipe dispenser
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Small folding cart (SBC152123)  |  Next: Small Lab Frame, Aluminum