Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-98934-97
Brass holder shown with neoprene and plastic holders (not included)
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-98934-97
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Viscometer Accessories
Share
|
Brass viscometer holder for Zeitfuchs viscometers
YO-98934-97
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
For holding Zeitfuchs
®
cross-arm glass capillary viscometers in constant-temperature viscosity baths. Brass holder includes a handle.
Specifications
Product Type
Viscometer Accessory
Material
Brass
Viscometer type
For Zeitfuchs® cross-arm
Brand
Zeitfuchs
Manufacturer number
9726-M98
Model
9726-M98
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
L'affaire est ketchup? Pas si simple que ça...les mesures de viscosité
In the Thick of Things: Measuring Viscosity
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Viscous Fluid Behavior
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Brass viscometer holder for Zeitfuchs viscometers
Zeitfuchs Viscometers
Zeitfuchs Accessories for Viscosity Baths
Zeitfuchs Material Testing & Physical Testing
Cannon Viscometer Accessories
Ubbelohde Viscometers
Ubbelohde Type Viscometer
Ubbelohde Accessories for Viscosity Baths
Ubbelohde Material Testing & Physical Testing
Ubbelohde Physical Property Testing
Cannon Zeitfuchs Cross Arm Viscometers
Related Products for Brass viscometer holder for Zeitfuchs viscometers
Image
Product Detail
SKU
Neoprene viscometer holder for Zeitfuchs viscometers
Category: Viscometer Accessories
Description: Accessories for Viscosity Baths
Brand:
Zeitfuchs
Model Number:
9726-M95
YO-98934-96
More Details
Brass viscometer holder for Ubbelohde viscometers (9726-M82)
Category: Viscometer Accessories
Description: Accessories for Viscosity Baths
Brand:
Ubbelohde
Model Number:
9726-M82
YO-98934-95
More Details
Brass viscometer holder for Cannon-Ubbelohde viscometers
Category: Viscometer Accessories
Description: Accessories for Viscosity Baths
Brand:
Cannon
Model Number:
9726-M76
YO-98934-94
More Details
Brass viscometer holder for all Cannon-Fenske type viscometers
Category: Viscometer Accessories
Description: Accessories for Viscosity Baths
Brand:
Cannon
YO-98934-98
More Details
Cannon Zeitfuchs cross-arm viscometer, size 1
Category: Cannon Glass Capillary Viscometers
Description: Zeitfuchs Cross-Arm Viscometer
Brand:
Cannon
Model Number:
9723-S50
YO-98934-00
More Details
Cannon Zeitfuchs cross-arm viscometer, size 2
Category: Cannon Glass Capillary Viscometers
Description: Zeitfuchs Cross-Arm Viscometer
Brand:
Cannon
Model Number:
9723-S53
YO-98934-01
More Details
Cannon Zeitfuchs cross-arm viscometer, size 3
Category: Cannon Glass Capillary Viscometers
Description: Zeitfuchs Cross-Arm Viscometer
Brand:
Cannon
Model Number:
9723-S56
YO-98934-02
More Details
Cannon Zeitfuchs cross-arm viscometer, size 4
Category: Cannon Glass Capillary Viscometers
Description: Zeitfuchs Cross-Arm Viscometer
Brand:
Cannon
Model Number:
9723-S59
YO-98934-03
More Details
Cannon Zeitfuchs cross-arm viscometer, size 5
Category: Cannon Glass Capillary Viscometers
Description: Zeitfuchs Cross-Arm Viscometer
Brand:
Cannon
Model Number:
9723-S62
YO-98934-04
More Details
Cannon Zeitfuchs cross-arm viscometer, size 6
Category: Cannon Glass Capillary Viscometers
Description: Zeitfuchs Cross-Arm Viscometer
Brand:
Cannon
Model Number:
9723-S65
YO-98934-05
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Brass viscometer holder for Zeitfuchs viscometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Brass viscometer holder for Ubbelohde viscometers (9726-M82)
|
Next: Brass, Two-Way, Direct-Acting Valve (Normally Closed), 1/4" NPT(F), 1/ (8262H22-24VDC)
31125329-p-content_2 31125329pcontent2