Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Soil Nutrient Test Kit Macronutrient Micronutrient and pH
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
MSDS
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-99031-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Soil Testing
Share
|
Soil Nutrient Test Kit; Macronutrient, Micronutrient, and pH (5010-01)
YO-99031-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Quick, visible color-change indicates results
Refillable reagents minimize product usage costs
Soil test kits simplify measuring the pH level, nutrients, or texture of your soil. Tests use standard reagents to produce color changes; compare results to color charts for concentration determination. Determine macronutrient, micronutrient, and pH levels. Tests for calcium, chloride, potassium, and sulfate are based on turbidity measurements.
What's included:
test reagents, a soil management handbook for determining lime and fertilizer requirements, and a pad of soil analysis report forms. Refill reagents are available to replenish your kitsee "Accessories" at right.
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Range
Aluminum: L-H (5-125 ppm)
Calcium: 150-2800 ppm
Chloride: 25-500 ppm
Humus(organic): L-H (1-1/2-8%)
Iron(ferric): 5-125 lbs/acre
Magnesium: L-H (5-150 ppm)
Manganese: L-H (4-40 ppm)
Nitrogen(ammonia): L-H (5-150 ppm)
Nitrogen(nitrate): 10-150 lbs/acre
Nitrogen(nitrite): 1-50 ppm
pH: 3.8-9.6 pH
Phosphorus: 10-200 lbs/acre
Potassium: 100-400 lbs/acre
Sulfate: 50-2000 ppm
Tests/kit
Aluminum: 50
Calcium: 50
Chloride: 50
Humus(organic): 50
Iron(ferric): 50
Magnesium: 50
Manganese: 50
Nitrogen(ammonia): 50
Nitrogen(nitrate): 50
Nitrogen(nitrite): 50
pH:100
Phosphorus: 50
Potassium: 50
Sulfate: 50
Model
5010-01
Manufacturer number
5010-01
Accessories
Qty
/
Price
Availability
Replacement reagents for deluxe soil chemistry test kit
YO-99031-51
EACH
Call for availability.
Add to Cart
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for Soil Nutrient Test Kit; Macronutrient, Micronutrient, and pH (5010-01)
Nutrient Test Kit
Thermo Scientific Nutrient Test Kits
Soil Ph Testing Kits
Soil Texture Testing
Soil Testing Kits
Lamotte Soil Test Kits
Lamotte Test Kits
Test Kits
Lamotte Turbidity Test Kit
Methylene Blue Test Kit for Microfilm
Related Products for Soil Nutrient Test Kit; Macronutrient, Micronutrient, and pH (5010-01)
Image
Product Detail
SKU
Soil Nutrient Test Kit; Macronutrient and pH (5029)
Category: Soil Testing
Description: LaMotte Soil Test Kits
Brand:
LaMotte
Model Number:
5029
YO-99031-00
More Details
Soil texture test kit (1067)
Category: Soil Testing
Description: LaMotte Soil Test Kits
Brand:
LaMotte
Model Number:
1067
YO-99030-00
More Details
Garden test kit (5679)
Category: Soil Testing
Description: LaMotte Soil Test Kits
Brand:
LaMotte
Model Number:
5679
YO-99030-10
More Details
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 40 ppb (K-7540)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Dissolved Oxygen Test Kits
Brand:
Titrets
Model Number:
K-7540
YO-05540-20
More Details
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 100 ppb (K-7599)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Dissolved Oxygen Test Kits
Brand:
Titrets
Model Number:
K-7599
YO-05540-25
More Details
CHEMets Colorimetric Dissolved Oxygen Test Kit, 0 to 1 ppm (K-7501)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Dissolved Oxygen Test Kits
Brand:
Titrets
Model Number:
K-7501
YO-05540-30
More Details
CHEMets Colorimetric Dissolved Oxygen Test Kit, 1 to 12 ppm (K-7512)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Dissolved Oxygen Test Kits
Brand:
Titrets
Model Number:
K-7512
YO-05540-40
More Details
Aqua Comparator Test Kit, Nitrate, 10-100 mg/L (L147400)
Category: pH Test Strips, pH Indicators, and Water Quality Testing Kits
Description: Aqua Comparator Test Kits
Brand:
Aqua Comparator
Model Number:
L147400
YO-05542-44
More Details
Colorimetric Chromate Test Kit (0554264)
Category: Water Quality Test Kits
Description: Cole-Parmer Colorimetric Test Kits
Brand:
Cole-Parmer
Model Number:
0554264
YO-05542-64
More Details
Colorimetric Phosphate, Ortho Test Kit (0554270)
Category: Water Quality Test Kits
Description: Cole-Parmer Colorimetric Test Kits
Brand:
Cole-Parmer
Model Number:
0554270
YO-05542-70
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Soil Nutrient Test Kit; Macronutrient, Micronutrient, and pH (5010-01)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Soil Nutrient Test Kit; Macronutrient and pH (5029)
|
Next: Soil pH Electrode, BNC Connector (YO-05992-62)
31125329-p-content_2 31125329pcontent2