Subtotal =  
Items =  
View Cart
Welcome
 
Home  >  Water Deionization Systems   

Thermo Scientific Barnstead E-pure™ 4-Holder Water Purification Sys (D4641)

YO-99252-10
Loading price...

Call for availability.
Qty

  • Produce ASTM Type I water
  • Recirculation pump ensures water purity
Thermo Scientific Barnstead E-pure systems meet or exceed ASTM, CAP, and NCCLS requirements for bacteria free, Type I reagent grade water. Inert wetted parts prevent contamination of purified water. A digital resistivity meter (included) indicates water purity. A built-in, whisper-quiet recirculation pump with pressure regulator ensures Type I reagent grade water on demand. Order low-water and low-pressure pump protectors below at right.
Choose from wall mountable three- or four-holder purification systems. Order disposable cartridges (required) by searching our Replacement Cartridge section; upgrade your system by adding an optional Remote Dispenser.
Product Type Water Purification System
Max flow rate 2 liters per minute
Inlet pressure (min) 5 psi or gravity feed
Inlet pressure (Max) 100 psi
Inlet port 1/4" NPT(F)
Inlet water temperature 40 to 120F (4 to 49C)
No. of holders 4
Dimensions 35"W x 28"H x 7-1/2"D
VAC 120
Model D4641
Brand E-pure
Manufacturer number D4641

Accessories
Qty/Price
Availability

EACH

Call for availability.

EACH

Call for availability.
Build a complete system




Technical Resources
 
Related Products for Thermo Scientific Barnstead E-pure™ 4-Holder Water Purification Sys (D4641)
Image
  Product Detail
SKU

Thermo Scientific Barnstead E-pure™ 4-Holder Water Purification Sys (D4642-33)

Category: Water Deionization Systems
Description: Thermo Scientific Barnstead E-pure 3- and 4-Holder Water Purification Systems
Brand: E-pure
Model Number: D4642-33


Thermo Scientific Barnstead E-pure™ 3-Holder Water Purification Sys (D4631)

Category: Water Deionization Systems
Description: Thermo Scientific Barnstead E-pure 3- and 4-Holder Water Purification Systems
Brand: E-pure
Model Number: D4631


Thermo Scientific Barnstead E-pure™ 3-Holder Water Purification Sys (D4632-33)

Category: Water Deionization Systems
Description: Thermo Scientific Barnstead E-pure 3- and 4-Holder Water Purification Systems
Brand: E-pure
Model Number: D4632-33


High-Purity Cartridge for Thermo Scientific Barnstead Mega-Pure Distil

Category: Mega-Pure Water Purification Stills
Description: Cartridges for Thermo Scientific Barnstead Mega-Pure Distillation Systems Pretreatment Deionizers
Brand: Thermo Scientific Barnstead
Model Number: D400377


Thermo Scientific Barnstead Single-Holder System (D4511)

Category: B-pure Water Purification Systems
Description: Thermo Scientific Barnstead B-Pure Single-Holder System
Brand: B-pure
Model Number: D4511


Thermo Scientific Barnstead Double-Holder System, 120 VAC (D4521)

Category: B-pure Water Purification Systems
Description: Thermo Scientific Barnstead B-Pure Double-Holder System
Brand: B-pure
Model Number: D4521


Thermo Scientific Barnstead Double-Holder System, 230 VAC (D4522-33)

Category: B-pure Water Purification Systems
Description: Thermo Scientific Barnstead B-Pure Double-Holder System
Brand: B-pure
Model Number: D4522-33


Thermo Scientific Barnstead Mega-Pure Glass Still, 1.0 LPH, 120 VAC

Category: Mega-Pure Water Purification Stills
Description: Thermo Scientific Barnstead Mega-Pure Glass Stills
Brand: Mega-Pure
Model Number: A440267


Thermo Scientific Barnstead Mega-Pure Glass Still, 1.0 LPH, 240 VAC

Category: Mega-Pure Water Purification Stills
Description: Thermo Scientific Barnstead Mega-Pure Glass Stills
Brand: Mega-Pure
Model Number: A7982


Thermo Scientific Barnstead Mega-Pure Glass Still, 3.4 LPH, 208 VAC

Category: Mega-Pure Water Purification Stills
Description: Thermo Scientific Barnstead Mega-Pure Glass Stills
Brand: Mega-Pure
Model Number: A440696


Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews