Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Hach 2100N Laboratory Turbidity Meter EPA compliant 230 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-99512-07
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Turbidity Benchtop Meters
Share
|
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 230 VAC
YO-99512-07
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Patented ratio optical system ensures accurate, stable readings
Smart self-diagnostics alert you to problems including using the wrong calibration standard
Meets U.S. EPA method 180.1
Wide measuring range using three selectable readout modes0 to 4000 nephelometric turbidity units, 0 to 26,800 NEPH (Nephelos), and 0 to 980 EBC (European Brewing Convention)
Optional ratioing switches instrument from multidetector ratio mode to nonratio
Signal averaging minimizes noise and display fluctuations
Air purge system connection purges the optical compartment with dry air and minimizes condensation on the sample cell
These benchtop turbidity meters offer the most versatile and advanced instruments available for nephelometric measurement. Push-button calibration and operation eliminates manual adjustments or zeroing. The patented optical system provides accurate results over a wide range of samples and turbidity, simplifying routine laboratory work.
What's Included:
six sample cells; 30 mL each of <0.1, 20, 200, 1000, and 4000 NTU StablCal
®
stabilized formazin primary standards; silicone oil and oiling cloth; dust cover; and power cord.
Specifications
Product Type
Turbidity Meter
Meter style
Benchtop
Dimensions (" W)
15-3/4
Dimensions (" H)
5-5/8
Dimensions (" L)
12
Range (Turbidity)
0 to 4000
Resolution (Turbidity)
0.01, 0.1, 1
Accuracy (Turbidity)
± 2% of reading
Display
LED
Interface
RS-232
Power
230 VAC
CE Compliance
Yes
Brand
Hach
Model
2100N
Accessories
Qty
/
Price
Availability
Calibration sets for Hach Laboratory Turbidimeter, item numbers 99512-00, -05
YO-99512-50
EACH
Call for availability.
Add to Cart
Degassing kit
YO-99512-68
EACH
Call for availability.
Add to Cart
Sample cells, package of 6
YO-99512-70
EACH
Call for availability.
Add to Cart
Sample cell oiling cloth
YO-99512-72
EACH
Call for availability.
Add to Cart
Replacement silicone oil, 15-mL.
YO-99512-75
EACH
Call for availability.
Add to Cart
Sample cell adapter, 19 mm
YO-99512-78
EACH
Call for availability.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Good Laboratory Practices for Taking Turbidity Measurements
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Related Searches for Hach 2100N Laboratory Turbidity Meter, EPA compliant, 230 VAC
Hach 2100n Laboratory Turbidity Meters
Hach 2100an Laboratory Turbidity Meters
Laboratory Turbidity Meters
Hach Portable Turbidity Meter Kuwait
Turbidity Meter Hach
Laboratory Ph Meter Reviews
Turbidity Meters
Nephelometric Turbidity Meter
Hach Free Chlorine Benchtop Meters
Turbidity Meters and Turbidity Testing Products
Related Products for Hach 2100N Laboratory Turbidity Meter, EPA compliant, 230 VAC
Image
Product Detail
SKU
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 110 VAC
Category: Turbidity Benchtop Meters
Description: Hach 2100N Laboratory Turbidity Meters
Brand:
Hach
Model Number:
2100N
YO-99512-00
More Details
Hach 2100AN Laboratory Turbidity Meter, EPA compliant, 110 VAC
Category: Turbidity Benchtop Meters
Description: Hach 2100AN Laboratory Turbidity Meters
Brand:
Hach
YO-99512-10
More Details
Hach 2100AN IS Laboratory Turbidity Meter, ISO compliant, 230 VAC
Category: Turbidity Benchtop Meters
Description: Hach 2100AN Laboratory Turbidity Meters
Brand:
Hach
Model Number:
2100AN IS
YO-99512-15
More Details
Hach 2100Q Handheld Turbidity Meter, EPA compliant
Category: Turbidity Handheld Meters
Description: Hach 2100Q Handheld Turbidity Meter
Brand:
Hach
Model Number:
2100Q Portable Turbidity Meter
YO-99511-10
More Details
Cole-Parmer Field Benchtop Turbidity Meters, EPA compliant, 110 VAC
Category: Turbidity Benchtop Meters
Description: Cole-Parmer Field Benchtop Turbidity Meters
Brand:
Cole-Parmer
Model Number:
11520-110
YO-08391-50
More Details
Cole-Parmer Field Benchtop Turbidity Meters, EPA compliant, 220 VAC
Category: Turbidity Benchtop Meters
Description: Cole-Parmer Field Benchtop Turbidity Meters
Brand:
Cole-Parmer
Model Number:
11520-220
YO-08391-55
More Details
Cole-Parmer Laboratory Benchtop Turbidity Meters, EPA compliant, 110 V
Category: Turbidity Benchtop Meters
Description: Cole-Parmer Laboratory Benchtop Turbidity Meters
Brand:
Cole-Parmer
Model Number:
11150-110V
YO-08391-40
More Details
Cole-Parmer Laboratory Benchtop Turbidity Meters, EPA compliant, 220 V
Category: Turbidity Benchtop Meters
Description: Cole-Parmer Laboratory Benchtop Turbidity Meters
Brand:
Cole-Parmer
Model Number:
11150-220V
YO-08391-45
More Details
LaMotte 2020we Handheld Turbidity Meter, EPA compliant
Category: Turbidity Handheld Meters
Description: LaMotte 2020 Handheld Turbidity Meters
Brand:
LaMotte
Model Number:
1970-EPA
YO-05563-22
More Details
HF Scientific Micro 100 Benchtop Turbidity Meter, EPA compliant, 110 V
Category: Turbidity Benchtop Meters
Description: HF Scientific Micro 100 Benchtop Turbidity Meters
Brand:
HF Scientific
Model Number:
20001
YO-99543-00
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 230 VAC
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hach 2100N Laboratory Turbidity Meter, EPA compliant, 110 VAC
|
Next: Hach 2100Q Handheld Turbidity Meter, EPA compliant
31125329-p-content_2 31125329pcontent2