Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-99574-04
Hach
®
Test Kits
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-99574-04
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Colorimetry Reagents
Share
|
Hach Free Chlorine (USEPA) Test Kit (21055-69)
YO-99574-04
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Environmental test kits for your Hach Colorimeter
Order the test kits for the Hach colorimeter to complete your water testing protocol. More test kits are availableif you don't see the one you need, call our Application Specialists for availability at 800-323-4340 x 6012.
All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120VAC 0r 99574-65, 240 VAC shown below).
Specifications
Product Type
Water Testing Kit
Tests/kit
100
Range
0 to 2.0 ppm
Manufacturer number
21055-69
Brand
Hach
Model
21055-69
Accessories
Qty
/
Price
Availability
Hach DR/870 Colorimeter
YO-99573-00
EACH
Call for availability.
Add to Cart
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Hach Free Chlorine (USEPA) Test Kit (21055-69)
Hach Free Chlorine Benchtop Meters
Chlorine Water Test Kit
Hach Dissolved Oxygen Water Test Kit
Hach Test Kits
Chlorine Testing Kits
Free Water Testing Kits
Chlorine and Ph Test Kit
Hach Free Reagents
Water Nitrates Test Kit
Iron Bacteria Test Kits
Related Products for Hach Free Chlorine (USEPA) Test Kit (21055-69)
Image
Product Detail
SKU
Hach Total Chlorine (USEPA) Test Kit (21056-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21056-69
YO-99574-05
More Details
Hach Total Iron (USEPA) Test Kit (21057-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21057-69
YO-99574-09
More Details
Hach Low-range Nitrite (USEPA) Test Kit (21071-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21071-69
YO-99574-13
More Details
Hach Phosphorus (USEPA) Test Kit (21060-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21060-69
YO-99574-17
More Details
Hach Sulfate (USEPA) Test Kit (21067-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21067-69
YO-99574-21
More Details
Hach High-range Nitrate Test Kit (21061-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21061-69
YO-99574-11
More Details
Hach Manganese (USEPA) Test Kit (24300-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
24300-00
YO-99574-10
More Details
Hach Sulfide (USEPA) Test Kit (22445-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
22445-00
YO-99574-22
More Details
Hach Low-range Nitrate Test Kit (24298-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
24298-00
YO-99574-12
More Details
Hach Total Nitrogen (TNT) Test Kit (26722-45)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
26722-45
YO-99574-14
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hach Free Chlorine (USEPA) Test Kit (21055-69)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hach Fluoride (SPADNS) Reagent
|
Next: Hach Free Copper Test Kit (26033-00)
31125329-p-content_2 31125329pcontent2