Home  >  Colorimetry Reagents >
  

Hach Fluoride (SPADNS) Reagent

YO-99574-08
Loading price...

Call for availability.
Qty

  • Environmental test kits for your Hach Colorimeter
Order the test kits for the Hach colorimeter to complete your water testing protocol. More test kits are available—if you don't see the one you need, call our Application Specialists for availability at 800-323-4340 x 6012.
All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120VAC 0r 99574-65, 240 VAC shown below).

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Water Testing Kit
Tests/kit 25
Range 0 to 2.0 ppm
Model 25060-25
Manufacturer number 25060-25
Brand HACH





Technical Resources
 
Related Products for Hach Fluoride (SPADNS) Reagent
Image
  Product Detail
SKU

Ammonium Fluoride, Reagent ACS (5g) (AC423290050)

Category: Ammonium
Description: Ammonium Fluoride
Brand: Cole-Parmer
Model Number: AC423290050


p-Toluenesulfonyl fluoride 97% (5g) (AC152380050)

Category: Thymol to Toluene
Description: p-Toluenesulfonyl fluoride
Brand: Cole-Parmer
Model Number: AC152380050


Hydrogen fluoride-pyridine 65-70% (100g) (AC180071000)

Category: Homatro to Hydroq
Description: Hydrogen fluoride-pyridine
Brand: Cole-Parmer
Model Number: AC180071000


α-Toluenesulfonyl fluoride 99% (1g)

Category: Thymol to Toluene
Description: α-Toluenesulfonyl fluoride
Brand: Cole-Parmer
Model Number: AC215740010


α-Toluenesulfonyl fluoride 99% (5g)

Category: Thymol to Toluene
Description: α-Toluenesulfonyl fluoride
Brand: Cole-Parmer
Model Number: AC215740050


α-Toluenesulfonyl fluoride 99% (10g)

Category: Thymol to Toluene
Description: α-Toluenesulfonyl fluoride
Brand: Cole-Parmer
Model Number: AC215740100


Hach Aluminum Test Kit

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH
Model Number: 22420-00


Hach Free Chlorine (USEPA) Test Kit (21055-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH
Model Number: 21055-69


Hach Total Chlorine (USEPA) Test Kit (21056-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH
Model Number: 21056-69


Hach Free Copper Test Kit (26033-00)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH
Model Number: 26033-00


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

3 Questions | 3 Answers
Displaying questions 1-3
  • Q:

    is there 25 testing strips in one package?
    Asked on 2/18/2011 by albert from miami fl

    1 answer

    • STAFF EXPERT

      A:

      You will have everything you need in order to do 25 tests with each of these test kits. All reagents in the test kit are packaged as powder pillows.
      These test kits are meant to be used with the Hach DR/870 Colorimeter (99573-00). This colorimeter does not use test strips.

      Answered on 2/19/2011 by Jaime from Technical Support
  • Q:

    can i use this kit for finding the fluoride content in pastes.I have a colorimeter.
    Asked on 1/13/2011 by lalu from kerala

    1 answer

    • Staff Reviewer

      A:

      This kit has only been used to test water based solutions. If you take a small portion or paste and mix it into water to complete the sample volume this may work for you. However, we do not have an exact method or amount to mix in our suggest.

      Answered on 1/14/2011 by Jen from Technical Support
  • Q:

    I am looking for a home tap water test kit to measure levels of fluoride and heavy metals. Is this the right one?
    Asked on 7/29/2010 by Shoshone

    1 answer

    • STAFF EXPERT

      A:

      This is the kit that contains the test reagents required for colorimetric testing of water for fluoride. To take the measurements one will require a colorimeter such as the 99573-00.

      Answered on 8/2/2010 by Josh from Technical Support
Displaying questions 1-3

Do you have a question about this product? 

Product Reviews

Hach Fluoride (SPADNS) Reagent
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Hach DR/870 Colorimeter  |  Next: Hach Formaldehyde Drop Test Kit