Hach 26722-45 Colorimeter Test Kit , Total Nitrogen (Tnt), 0 to 25 ppm, 50/Kit
Representative image only.
Hach Mfr # 26722-45 – Item # EW-99574-14
Environmental test kits for your Hach Colorimeter
  • All reagents in kits are packaged as powder pillows
  • 1 year warranty
PRODUCT OPTIONS
$245.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersTotal Nitrogen (TNT)
  • Number of Tests Included50
  • Range0 to 25 ppm
  • DescriptionColorimeter Test Kit , Total Nitrogen (Tnt), 0 to 25 ppm, 50/Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • All reagents in kits are packaged as powder pillows
  • 1 year warranty
More About this Item
Order the test kits for the Hach colorimeter to complete your water testing protocol. All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120 VAC or 99574-65, 240 VAC).
Requires COD reactor listed below.