Home  >  Colorimetry Reagents >
  

Hach Total Phosphorus(USEPA) Test Kit (27426-45)

YO-99574-18
Loading price...

Call for availability.
Qty

  • Environmental test kits for your Hach Colorimeter
Order the test kits for the Hach colorimeter to complete your water testing protocol. More test kits are available—if you don't see the one you need, call our Application Specialists for availability at 800-323-4340 x 6012.
All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120VAC 0r 99574-65, 240 VAC shown below).
Requires COD reactor listed below.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Water Testing Kit
Tests/kit 50
Range 0 to 3.5 ppm
Model 27426-45
Manufacturer number 27426-45
Brand HACH

Accessories
Qty/Price
Availability

EACH

Call for availability.

EACH

Call for availability.
Test tube rack
YO-99574-54

EACH

Call for availability.

EACH

Usually ships in 14 days or sooner.

EACH

Call for availability.




Technical Resources
 
Related Products for Hach Total Phosphorus(USEPA) Test Kit (27426-45)
Image
  Product Detail
SKU

Hach Total Nitrogen (TNT) Test Kit (26722-45)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH


Hach Total Chlorine (USEPA) Test Kit (21056-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH


Hach Total Iron (USEPA) Test Kit (21057-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH


Hach Phosphorus (USEPA) Test Kit (21060-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH


Hach Free Chlorine (USEPA) Test Kit (21055-69)

Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand: HACH


Hach Phosphate, Total Color Disc Test Kits

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach Chlorine, Free and Total Color Disc Test Kit

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach Chlorine, Total Color Disc Test Kit (YO-05545-70)

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach Chlorine Free and Total Color Disc Test Kit

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach Copper, Free and Total Color Disc Test Kit

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    I am looking for a TP test we can use in the field. Does this kit require the COD reactor?
    Asked on 1/19/2010 by Mary from Milwaukee, WI

    1 answer

    • Staff Reviewer

      A:

      Yes, this test requires a COD reactor like 99574-60.

      Answered on 10/29/2010 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews
Customers Who Bought This Item Also Bought

Hach Total Phosphorus(USEPA) Test Kit (27426-45)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Hach Total Nitrogen (TNT) Test Kit (26722-45)  |  Next: Hach Waterproof ISFET Multiparameter Meter with Probes