Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Hach COD Test Kit High Range 0 to 1500 ppm
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-99574-26
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH/Ion Meter Accessories
Share
|
Hach COD Test Kit, High Range, 0 to 1500 ppm
YO-99574-26
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Kits contain leak-proof vials. Caps provide a positive seal that traps volatile material and ensures complete digestion. Reagents are premixed, minimizing dangerous spills and handling of corrosive and toxic chemicals. Small reagent volumes mean less silver and mercury waste. Pack of 25 reagent vials.
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Accessories
Qty
/
Price
Availability
Hach DR/870 Colorimeter
YO-99573-00
EACH
Call for availability.
Add to Cart
Volumetric pipette, 2.00 mL
YO-99574-52
EACH
Call for availability.
Add to Cart
Pipette filler, safety bulb
YO-99574-53
EACH
Call for availability.
Add to Cart
Test tube rack
YO-99574-54
EACH
Call for availability.
Add to Cart
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Hach COD Test Kit, High Range, 0 to 1500 ppm
Hach Water Testing Kits
Hach Colorimetry Test Kits
Cod Test
Bel Art Colorimeter Test Kits
Ozone Test Kits
Pocket Colorimeter Ii Test Kit
Hach Test Kits
Water Nitrates Test Kit
Cole-Parmer Arsenic Test Kit
Dissolved Oxygen Test Kit Test Kits
Related Products for Hach COD Test Kit, High Range, 0 to 1500 ppm
Image
Product Detail
SKU
YSI COD Test Kits, High-range, 25 Vials
Category: Chemical Oxygen Demand (COD) Test Systems
Description: YSI 910 COD Reactor, Test Kits, and Colorimeter
Brand:
YSI
YO-99550-62
More Details
Hach COD Test Kit, Low Range, 0 to 150 ppm
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Hach
YO-99574-25
More Details
YSI COD Test Kits, Low-range, 25 Vials
Category: Chemical Oxygen Demand (COD) Test Systems
Description: YSI 910 COD Reactor, Test Kits, and Colorimeter
Brand:
YSI
YO-99550-56
More Details
YSI COD Test Kits, Medium-range, 25 Vials
Category: Chemical Oxygen Demand (COD) Test Systems
Description: YSI 910 COD Reactor, Test Kits, and Colorimeter
Brand:
YSI
YO-99550-58
More Details
Hach Aluminum Test Kit
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
22420-00
YO-99574-03
More Details
Hach Cyanide Test Kit (24302-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
24302-00
YO-99574-07
More Details
Hach Ozone Test Kit (25170-25)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
25170-25
YO-99574-16
More Details
Hach Zinc Test Kit (24293-00)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
24293-00
YO-99574-23
More Details
Hach TOC Test Kit, High Range, 20 to 700 mg/L
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Hach
YO-99574-29
More Details
Hach High-range Nitrate Test Kit (21061-69)
Category: Colorimetry Reagents
Description: Hach Colorimetry Test Kits
Brand:
Hach
Model Number:
21061-69
YO-99574-11
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hach COD Test Kit, High Range, 0 to 1500 ppm
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hach COD reactor, 220 VAC, 50/60Hz
|
Next: Hach COD Test Kit, Low Range, 0 to 150 ppm
31125329-p-content_2 31125329pcontent2