Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Cole Parmer One Shot conductivity standards 100 S 100 mL vials 6 PK
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
More info
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-00652-64
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Conductivity Calibration Solutions
Share
|
Cole-Parmer One-Shot™ conductivity standards, 100 µS, 100 mL via
YO-00652-64
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
NIST-traceable reference materials include a calibration report from an ISO Guide 34 and ISO 17025 accredited laboratory for the highest accuracy available—up to ±0.25%!
These convenient single-use vials eliminate contamination concerns and are ideal for the lab as well as the field. Calibration is performed in the large 3-1⁄2"H x 1-3⁄4" dia, PE vial that accommodates virtually any conductivity probe. Includes a NIST-traceable calibration report supplied by the manufacturer and temperature compensation chart. Pack of six vials, each containing 100 mL of conductivity solution.
Specifications
Product Type
Calibration Solutions
Conductivity
100 µS
Size
100 mL vials, 6/PK
Brand
Cole-Parmer
Model
4176CP
Manufacturer number
4176CP
Technical Resources
MSDS 1 (76 KB)
Cole-Parmer Traceable
Conductivity Standards (270 KB)
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
More info
Sample of certificate of analysis
Related technical articles (Roll over for summary)
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show Less...
Related Searches for Cole-Parmer One-Shot™ conductivity standards, 100 µS, 100 mL via
One Shot Conductivity Calibration Standards
Cole-Parmer Calibration Products and Standards
Cole-Parmer Round Panel Jacks - Standard
Cole-Parmer One-Way Luer Check Valves
Cole-Parmer One-Tier Mills
Cole-Parmer Rectangular Mounting Panels - Standard
Cole-Parmer One and Two-Channel Flatbed Recorders
Cole-Parmer Traceable Portable Conductivity Meters
Cole-Parmer Correlated Flowmeters with Standard Valves
Cole-Parmer Twist-Lock Conductivity Sensors
Related Products for Cole-Parmer One-Shot™ conductivity standards, 100 µS, 100 mL via
Image
Product Detail
SKU
Cole-Parmer One-Shot™ conductivity standard assortment pack, 100 mL
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4172CP
YO-00652-74
More Details
Cole-Parmer One-Shot™ conductivity standards, 5 µS, 100 mL vials
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4271CP
YO-00652-60
More Details
Cole-Parmer One-Shot™ conductivity standards, 10 µS, 100 mL vial
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4175CP
YO-00652-62
More Details
Cole-Parmer One-Shot™ conductivity standards, 1000 µS, 100 mL vi
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4177CP
YO-00652-66
More Details
Cole-Parmer One-Shot™ conductivity standards, 1413 µS, 100 mL vi
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4174CP
YO-00652-68
More Details
Cole-Parmer One-Shot™ conductivity standards, 10,000 µS, 100 mL
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4178CP
YO-00652-70
More Details
Cole-Parmer One-Shot™ conductivity standards, 100,000 µS, 100 mL
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
Model Number:
4179CP
YO-00652-72
More Details
Cole-Parmer One-Shot™ conductivity standards, 150,000 µS, 100 mL
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00652-76
More Details
Cole-Parmer One-Shot™ conductivity standards, 200,000 µS, 100 mL
Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand:
Cole-Parmer
YO-00652-78
More Details
One-Shot™ conductivity standards, 150,000 µS, 100 mL vials, 6/PK
Category: Conductivity Calibration Solutions
Description: Control Company Traceable One-Shot Conductivity Solutions with NIST/ISO 34, ISO 17025/A2LA Certificate
Brand:
Control Company
YO-00652-49
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Cole-Parmer One-Shot™ conductivity standards, 100 µS, 100 mL via
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Cole-Parmer One-Shot™ conductivity standards, 10,000 µS, 100 mL
|
Next: Cole-Parmer One-Shot™ conductivity standards, 100,000 µS, 100 mL
31125329-p-content_2 31125329pcontent2