Cole-Parmer One-Shot ™ conductivity standards, 100 µS, 100 mL vials, 6/PK

YO-00652-64
Loading price...

Usually ships in 14 days or sooner.
Qty

  • NIST-traceable reference materials include a calibration report from an ISO Guide 34 and ISO 17025 accredited laboratory for the highest accuracy available—up to ±0.25%!
These convenient single-use vials eliminate contamination concerns and are ideal for the lab as well as the field. Calibration is performed in the large 3-1⁄2"H x 1-3⁄4" dia, PE vial that accommodates virtually any conductivity probe. Includes a NIST-traceable calibration report supplied by the manufacturer and temperature compensation chart. Pack of six vials, each containing 100 mL of conductivity solution.
Product Type Conductivity Standard
Conductivity 100 µS
Size 100 mL vials, 6/PK
Brand Cole-Parmer
Model 4176CP
Manufacturer number 4176CP





Technical Resources
 
Related Products for Cole-Parmer One-Shot ™ conductivity standards, 100 µS, 100 mL vials, 6/PK
Image
  Product Detail
SKU

Cole-Parmer One-Shot Conductivity Solution Assortment; 100 mL

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot ™ conductivity standards, 5 µS, 100 mL vials, 6/PK

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot ™ conductivity standards, 10 µS, 100 mL vials, 6/PK

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot ™ conductivity standards, 1000 µS, 100 mL vials, 6/PK

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot ™ conductivity standards, 1413 µS, 100 mL vials, 6/PK

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot 10,000 uS Conductivity Solution; 100 mL/Vial,

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot 100,000 uS Conductivity Solution; 100 mL/Vial,

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot 150,000 uS Conductivity Solution; 100 mL/Vial, 6/Pk

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


Cole-Parmer One-Shot 200,000 uS Conductivity Solution; 100 mL/Vial, 6/Pk

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions, NIST-Traceable Reference Materials
Brand: Cole-Parmer


One-Shot ™ conductivity standards, 150,000 µS, 100 mL vials, 6/PK

Category: Conductivity Calibration Solutions
Description: Cole-Parmer One-Shot Conductivity Solutions with NIST/ISO 34, ISO 17025/A2LA Certificate
Brand: Control Company


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought