Thermo Scientific Barnstead Pretreatment Cartridge (D0835)

YO-01505-22
Loading price...

Usually ships in 14 days or sooner.
Qty
Product Type Water Purification Cartridge
Capacity As determined by feed water quality
Compatible with All NANOpure™ E-pure™, B-pure™ systems (except Nanopure Infinity)
Capacity (Liters) 18,925
Function Removes colloids, chlorine, and organics; use when nonionized solids and/or colloids are present
Capacity (gal.) 5000
Manufacturer number D0835
Brand Thermo Scientific Barnstead





Technical Resources
 
Related Products for Thermo Scientific Barnstead Pretreatment Cartridge (D0835)
Image
  Product Detail
SKU

Thermo Scientific Barnstead Pretreatment cartridge, Pore size: 10 µm

Category: Water Purification Cartridges for Thermo Scientific Barnstead Half-Sized Holder System
Description: Thermo Scientific Barnstead Pretreatment cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment cartridge, Pore size: 15 µm

Category: Water Purification Cartridges for Thermo Scientific Barnstead Half-Sized Holder System
Description: Thermo Scientific Barnstead Pretreatment cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment cartridges, Pore size: 1 µm

Category: Water Purification Cartridges for Thermo Scientific Barnstead Half-Sized Holder System
Description: Thermo Scientific Barnstead Pretreatment cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment cartridge, Pore size: 5 µm

Category: Water Purification Cartridges for Thermo Scientific Barnstead Half-Sized Holder System
Description: Thermo Scientific Barnstead Pretreatment cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment Cartridge (D0836)

Category: Water Purification Cartridges
Description: Thermo Scientific Barnstead Replacement Cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment Cartridge for reverse osmosis and distilled feedwater sources

Category: NANOpure Infinity Water Purification Systems Cartridges
Description: Thermo Scientific Barnstead Replacement Cartridges for NANOpure Infinity Systems
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment Cartridge for tap and deionized feedwater sources

Category: NANOpure Infinity Water Purification Systems Cartridges
Description: Thermo Scientific Barnstead Replacement Cartridges for NANOpure Infinity Systems
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment Cartridge (D0813)

Category: Water Purification Cartridges
Description: Thermo Scientific Barnstead Replacement Cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Pretreatment Cartridge (D0832)

Category: Water Purification Cartridges
Description: Thermo Scientific Barnstead Replacement Cartridges
Brand: Thermo Scientific Barnstead


Thermo Scientific Barnstead Organic-free cartridge, 330 grains as NaCl (D50252)

Category: NANOpure Infinity Water Purification Systems Cartridges
Description: Thermo Scientific Barnstead Replacement Cartridges for NANOpure Infinity Systems
Brand: Thermo Scientific Barnstead


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought

Thermo Scientific Barnstead Pretreatment Cartridge (D0835)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Thermo Scientific Barnstead Pretreatment Cartridge (D0832)  |  Next: Thermo Scientific Barnstead Pretreatment Cartridge (D0836)