Home  >  Fermentation Components & Accessories >
  

Chemostat tube for 15-L vessels. (YO-01919-46)

YO-01919-46
$3,135.38 AUD/EACH

Call for availability.
Qty
    Chemostat tubes are used with a Masterflex® tubing pump (sold separately) to draw off harvested media or to add fresh media to maintain constant media level.





    Technical Resources
     
    Related Products for Chemostat tube for 15-L vessels. (YO-01919-46)
    Image
      Product Detail
    SKU

    Chemostat tube for 2-, 3-, 5-, and 7-L vessels. (YO-01919-14)

    Category: Fermentation Components & Accessories
    Description: Chemostat tube
    Brand: Cole-Parmer


    Nipple holder for 15-L vessels (YO-01919-45)

    Category: Fermentation Components & Accessories
    Description: Nipple holder
    Brand: Cole-Parmer


    Heating blanket for 15 L vessels (YO-01919-88)

    Category: Fermentation Components & Accessories
    Description: Heating Blankets
    Brand: HVAC/R


    Nipple holder for 2-, 3-, 5-, and 7-L vessels. (YO-01919-11)

    Category: Fermentation Components & Accessories
    Description: Nipple holder
    Brand: Cole-Parmer


    Blind Caps; port size: 18 mm (M18 x 1.5 mm thread) (YO-01919-13)

    Category: Fermentation Accessories and Replacement Parts
    Description: Blind Caps
    Brand: Cole-Parmer


    Heating blanket for 3 L vessels (YO-01919-85)

    Category: Fermentation Components & Accessories
    Description: Heating Blankets
    Brand: HVAC/R


    Heating blanket for 5 L vessels (YO-01919-86)

    Category: Fermentation Components & Accessories
    Description: Heating Blankets
    Brand: HVAC/R


    Heating blanket for 7 L vessels (YO-01919-87)

    Category: Fermentation Components & Accessories
    Description: Heating Blankets
    Brand: HVAC/R


    Centrifuge tube rack for 15-mL centrifuge tubes (YO-06696-02)

    Category: Tube Racks
    Description: 15-mL Centrifuge Tube Racks
    Brand: Guaranteed In Stock Program


    Masterflex High-Pressure PharMed BPT Tubing, L/S 15HP, 25' (YO-95664-15)

    Category: L/S High-Pressure Tubing for Masterflex L/S Pumps
    Description: L/S High-Pressure PharMed BPT Tubing for Masterflex L/S Pumps
    Brand: Masterflex


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews