Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific Nunc Immunoplates surface Maxisorp well shape round well volume 0 3mL
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-01928-08
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
96-Well Plates
Share
|
Thermo Scientific Nunc™ Immunoplates; surface, Maxisorp; well shape
YO-01928-08
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Four surfaces with different degree of hydrophilicity, ideal for solid phase immunoassays (ELISA)
Engineered for easy/reproducible passive absorption
Certified homogeneity of binding
The PolySorp surface is hydrophobic for binding biomolecules that have hydrophobic properties. New MediSorp is slightly hydrophilic and binds proteins with a mixed hydrophilic/hydrophobic composition. This special surface provides an ideal balance of high protein absorption for coating yet low assay non-specific binding. MaxiSorp is still more hydrophilic. It is optimized for binding immunoglobulin but also binds a wide range of other proteins. New MultiSorp is the most hydrophilic surface offered. It binds biomolecules with a hydrophilic composition such a glycoproteins and glycans. Standard 96-well format/ 86 x 128 mm.
Specifications
Product Type
Microplates
Surface treatment
MaxiSorp®
Well shape
Round
Well volume (mL)
0.3
Brand
Thermo Scientific Nunc
Manufacturer number
449824
Technical Resources
Related technical articles (Roll over for summary)
Effects of Mounting a Breathable Membrane on the 384-Well Plate
Isolating Genomic DNA from Whole Blood
Show Less...
Related Searches for Thermo Scientific Nunc™ Immunoplates; surface, Maxisorp; well shape
Thermo Scientific Nunc 96-Well Microplates
Thermo Scientific Nunc Microplate Products
Thermo Scientific Nunc Nunc-Immuno Washers
Thermo Scientific Nunc Roller Bottles
Thermo Scientific Nunc Replacement Accessories
Thermo Scientific Nunc Cryotube Vials
Thermo Scientific Nunc Cryogenics
Thermo Scientific Nunc Cooling Equipment
Thermo Scientific Nunc Labware
Thermo Scientific Nunc Slides/Cover Slips
Related Products for Thermo Scientific Nunc™ Immunoplates; surface, Maxisorp; well shape
Image
Product Detail
SKU
Thermo Scientific Nunc™ Immunoplates; surface, nontreated, nonsteri (YO-01928-02)
Category: 96-Well Plates
Description: Thermo Scientific Nunc 96-Well Microplates, Immunology
Brand:
Thermo Scientific Nunc
YO-01928-02
More Details
Thermo Scientific Nunc™ Immunoplates; surface, nontreated, sterile; (YO-01928-04)
Category: 96-Well Plates
Description: Thermo Scientific Nunc 96-Well Microplates, Immunology
Brand:
Thermo Scientific Nunc
YO-01928-04
More Details
Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
Category: 96-Well Plates
Description: Thermo Scientific Nunc 96-Well Microplates, Immunology
Brand:
Thermo Scientific Nunc
YO-01928-22
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-19)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-19
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-21)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-21
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 48-well Multi (YO-01930-24)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-24
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 4-well MultiD (YO-01930-25)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-25
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 12-well Multi (YO-01930-29)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-29
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 24-well Multi (YO-01930-40)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-40
More Details
Thermo Scientific Nunc 4-well culture dish, nontreated, 120/case
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc Non-Treated Polystyrene Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01928-60
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific Nunc™ Immunoplates; surface, Maxisorp; well shape
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific Nunc™ EZFlip™ Conical-Bottom Sterile 15 (YO-17411-32)
|
Next: Thermo Scientific Nunc™ Immunoplates; surface, nontreated, nonsteri (YO-01928-02)
31125329-p-content_2 31125329pcontent2