Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific Nunc Thermo Scientific Nunclon 12 well MultiDish with lid PS sterile
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-01930-29
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Essentials and Accessories for Microbial Analysis
Share
|
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 12-well Multi (YO-01930-29)
YO-01930-29
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Nunclon certified surface ensures optimum cell attachment and growthuse for scale up, cloning, viral titration, and fusion
Tested for cell growth and plating efficiency to ensure consistent biological performance
Raised well rims reduce risk of cross contamination
Round well styles feature vented lids for gas exchange and recessed areas around wells that act as a water reservoir to reduce evaporation from wells. Dishes with 6, 12, 24, and 48-well formats include lids with condensation rings to minimize evaporation. The 12 and 48-well dishes have a standard 96-well footprint. Rectangular well styles offer a larger culture area. They also accept standard glass cover slips: 24 x 60mm for 4-well plate; and 22 x 22mm for 8 well plate. Radiation sterilized.
Specifications
Product Type
Culture Dishes/Plates
Well dimensions
21.0 mm dia
Working volume
0.7 to 1.8 mL
Culture area
3.5 cm
2
/well
No. of wells
12
Quantity
1/sleeve, 75/case
Dimensions
86 x 128 mm
Manufacturer number
150628
Brand
Thermo Scientific Nunc
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Aspects of Nunc MAXISORP® MICROWELL® Certification
Negative Edge Effect in MICROWELL® ELISA
Principles in Adsorption to Polystyrene
Show All...
Stability of Nunc-Immuno® Maxisorp® Surfaces
Sterilized Disposable Bioprocess Consumables
Cole-Parmer Introduces Enhancements to Life Science Offering
Effects of Mounting a Breathable Membrane on the 384-Well Plate
Isolating Genomic DNA from Whole Blood
Show Less...
Related Searches for Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 12-well Multi (YO-01930-29)
Thermo Scientific Nunc Microbial Analysis
Nunc Thermo S
Thermo Scientific Nunc Roller Bottles
Thermo Scientific Nunc 96-Well Microplates
Thermo Scientific Nunc Nunc-Immuno Washers
Thermo Scientific Nunc Labware
Thermo Scientific Nunc Microplate Products
Thermo Scientific Nunc Replacement Accessories
Thermo Scientific Nunc Cryotube Vials
Thermo Scientific Nunc Cryogenics
Related Products for Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 12-well Multi (YO-01930-29)
Image
Product Detail
SKU
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 48-well Multi (YO-01930-24)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-24
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 24-well Multi (YO-01930-40)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-40
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-19)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-19
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 6-well MultiD (YO-01930-21)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-21
More Details
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 4-well MultiD (YO-01930-25)
Category: Essentials and Accessories for Microbial Analysis
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Delta MultiDishes
Brand:
Thermo Scientific Nunc
YO-01930-25
More Details
Thermo Scientific Nunc™ Nunclon™ cell culture flask, 25 cm2, ang
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Cell Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01930-41
More Details
Thermo Scientific Nunc™ Nunclon™ cell culture flask, 80 cm2, st
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Cell Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01930-42
More Details
Thermo Scientific Nunc™ Nunclon™ cell culture flask, 175 cm2, s
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc Thermo Scientific Nunclon Cell Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01930-44
More Details
Thermo Scientific Nunc Nunclon™ TripleFlask, 500 cm2, straight neck (YO-01931-00)
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc TripleFlask Tissue Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01931-00
More Details
Thermo Scientific Nunc Nunclon™ TripleFlask, 500 cm2, straight neck (YO-01931-10)
Category: Thermo Scientific Nunc Cell Culture Flasks
Description: Thermo Scientific Nunc TripleFlask Tissue Culture Flasks
Brand:
Thermo Scientific Nunc
YO-01931-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 12-well Multi (YO-01930-29)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific Nunc™ Thermanox Coverslips, 60 mm L x 22 mm W; 50
|
Next: Thermo Scientific Nunc™ Thermo Scientific Nunclon™ 24-well Multi (YO-01930-40)
31125329-p-content_2 31125329pcontent2