Spectra/por 4 Membranes Flat sheets; 150 x 150 mm; 25/pack (132712)

YO-02900-46
Loading price...

Call for availability.
Qty

  • Available in a variety of grades and molecular weight cutoffs (MWCO)
  • Available in a variety of dimensions to meet your application needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
Membrane discs and sheets are used to separate sample and buffer chambers in dialysis systems. Packaged 50 discs per pack and 25 sheets per pack.MWCO of 12,000 to 14,000
Product Type Dialysis Flat Discs and Sheets
Material Regenerated Cellulose
Membrane MWCO (Daltons) 12000
MWCO (Dalton) 12000 to 14000
Dimensions (mm) 150 x 150 mm sheet
Qty/pk 25
Brand Spectra/Por
Manufacturer number 132712





Technical Resources
 
Related Products for Spectra/por 4 Membranes Flat sheets; 150 x 150 mm; 25/pack (132712)
Image
  Product Detail
SKU

Spectra/Por 4 Membranes; flat width, 10mm; diameter, 6.4mm; length, 100'

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Spectra/Por 4 Membranes; flat width, 25mm; diameter, 15.9mm; length, 100'

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Spectra/Por 4 Membranes; flat width, 32mm; diameter, 20.4mm; length, 100'

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Spectra/Por 4 Membranes; flat width, 45mm; diameter, 28.6mm; length, 100'

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Spectra/Por 4 Membranes; flat width, 75mm; diameter, 47.7mm; length, 50'

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Tubing
Brand: Spectra/Por


Spectra/Por 3 Membranes Flat sheets; 108 x 108 mm; 25/pack (132723)

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Spectra/Por 3 Regenerated Cellulose (RC) Dialysis Discs and Sheets
Brand: Spectra/Por


Spectra/Por 4 Dialysis Membrane Disc, 12000 to 14000 MWCO, 33 mm dia; pack of 50.

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Discs and Sheets
Brand: Spectra/Por


Spectra/Por 4 Dialysis Membrane Disc, 12000 to 14000 MWCO, 47 mm dia; pack of 50.

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Discs and Sheets
Brand: Spectra/Por


Sepectr/Por 1 Membranes Flat sheets; 240 x 240 mm; 25/pack (132677)

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Spectra/Por 1 Regenerated Cellulose (RC) Dialysis Discs and Sheets
Brand: Spectra/Por


Spectr/Por 2 Membranes Flat sheets; 240 x 240 mm; 25/pack (132686)

Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Spectra/Por 2 Regenerated Cellulose (RC) Dialysis Discs and Sheets
Brand: Spectra/Por


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews