Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Washer For Uhp76 Tie Rod
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-02910-84
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Advantec Stirred Cells
Share
|
Washer For Uhp76 Tie Rod (341331)
YO-02910-84
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Increase membrane life and speed sample processing
Use stirred cells for dewatering and concentrating proteins and other biological samples with ultrafiltration membranes. They are also recommended for cellulose membrane filtration of fluids with heavy particle burden. Pressurized air from an external source (70 psi/4.8 bar max) filters sample through the membrane. Use cells with cellulose membranes on or ultrafiltration discs. Magnetic stir rod/bar are mounted to cell top cap and are preset to clear membrane surface. Samples do not touch any metal surfaces.
Cells include a preset safety relief valve for pressure buildup, a 1/2" NPT(M) cell cap connection, adapters, and pressure tubing. All cells have polyacetal end caps and stir rod, PTFE stir bar, polypropylene support screen, and silicone O-ring.
Technical Resources
Related technical articles (Roll over for summary)
Laboratory Filtration Selection Guide
Science in the Small Dairy Industry: Milk Sediment Testing
Basics of Cartridge Filtration
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Washer For Uhp76 Tie Rod (341331)
Free Standing Lab Frame
Cole-Parmer Glass Stir Rods
Cole-Parmer Nylon Monofilament Tie-on Filter Bags
Avery-Dennison Cable Ties
Cole-Parmer Glass Rods
Cole-Parmer Barbed Design Cable Ties
Ptfe Spatulas & Stir Rods
Cole-Parmer Tie Mounts
96 Well Microplate Washer
Masterflex Thrust Washers
Related Products for Washer For Uhp76 Tie Rod (341331)
Image
Product Detail
SKU
Plastic Clamp Ties, White, 100/pack (YO-95615-00)
Category: Tubing Clamps, and Accessories
Description: Plastic Clamp Ties
Brand:
Cole-Parmer
YO-95615-00
More Details
Plastic Clamp Ties, Black, 100/pack (YO-95615-10)
Category: Tubing Clamps, and Accessories
Description: Plastic Clamp Ties
Brand:
Cole-Parmer
YO-95615-10
More Details
Undercounter Glassware Washer 115V (YO-98950-01)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-01
More Details
Undercounter, General-Purpose Washer, 230V (YO-98950-05)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-05
More Details
Freestanding Glassware Washer, 115V (YO-98950-11)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-11
More Details
Undercounter Spindle-ready Washer, 230V (YO-98950-20)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-20
More Details
Free Standing Spindle-Ready Washer, 230V
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-30
More Details
Free Standing Spindle-ready Washer, 230V (CLW-1285I-FS) (YO-98950-35)
Category: Glassware Washers
Description: Cole-Parmer
®
Laboratory Glassware Washers
Brand:
Cole-Parmer
YO-98950-35
More Details
Support Rod, 28”, threaded, Stainless steel
Category: Laboratory Frame/Stand Rods
Description: Stainless Steel Standard Laboratory Support Rods
Brand:
Troemner
YO-04344-10
More Details
Ties 7.5”black 1000/pk
Category: Electrical Cable Ties
Description: Solid Cable Ties
Brand:
Cole-Parmer
Model Number:
08388-0
YO-06830-44
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Washer For Uhp76 Tie Rod (341331)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Washdown Process Pump, Digital Drive, 115/230 VAC
|
Next: Washer, Sealing Assembly (301916)
31125329-p-content_2 31125329pcontent2