Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific Multi Blok Analog Dry Block Heater Base one block 115 VAC 60 Hz
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-03129-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Thermo Scientific Multi-Blok ® Dry Block Heaters and Blocks
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-36402-08
,
browse similar items
or
Contact us for more information.
Thermo Scientific Multi-Blok® Analog Dry Block Heater Base; one-bl
YO-03129-00
Loading price...
Discontinued
Qty
:
Save to shopping list
Temperature uniformity within ±0.5degC (±0.9degF)
Three-position switch—high, low, or off
Low-range between ambient and 60degC (140degF) and
high-range to 130degC (266degF)
These versatile heating systems uniformly heat a wide range of test tubes simultaneously. Systems offer different combination of blocks for your specific needs. A complete heater system consists of a standard heater base and heating blocks (sold separately). Each block heater base features a stepless thermostat dial marked from 0 to 10 for repeatable settings. A separate power switch with power-on light operates independently of the thermostat dial.
Metal-sheathed safety heaters ensure temperature uniformity and rapid heating. Heater bases have steel housing with baked enamel finish, and include 6-ft, three-wire power cord; 115 VAC models include a plug.
Specifications
Max blocks
One
Product Type
Dry Block Heaters-Analog
Watts
100
Max temperature (º F)
266
Power (VAC)
115
Max temperature (º C)
130
Dimensions
8 in W x 3 in H x 5 in D
Number of Blocks
1
Brand
Thermo Scientific
Manufacturer number
2050Q
Model
2050Q
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Thermo Scientific Multi-Blok® Analog Dry Block Heater Base; one-bl
Thermo Scientific Multi-Blok Dry Heater
Thermo Scientific Multi-Blok Heating Blocks
Dry Block Heaters
Cole-Parmer Stabletemp Dry Block Heaters (Analog)
Glas-Col Dry Block Heaters
Cole-Parmer Stabletemp Dry Block Heaters (Digital)
Thermo Scientific Multi-Position Stirrer
Thermo Scientific Multi Channel Pipette
Dry Block Heaters-Analog
Digital Dry Block Heater
Related Products for Thermo Scientific Multi-Blok® Analog Dry Block Heater Base; one-bl
Image
Product Detail
SKU
Thermo Scientific Multi-Blok Analog Dry Block Heater Base; one-block,
Category: Thermo Scientific Multi-Blok
®
Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok
®
Analog Dry Block Heaters
Brand:
Multi-Blok
Model Number:
2050-1CEQ
YO-03129-05
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 1-block, 12
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
Model Number:
100
YO-36402-08
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 1-block, 23
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
YO-36402-09
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 2-block, 12
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
Model Number:
150
YO-36402-10
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 2-block, 23
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
YO-36402-11
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 3-block, 12
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
Model Number:
200
YO-36402-12
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 3-block, 23
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
YO-36402-13
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 4-block, 12
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
Model Number:
300
YO-36402-14
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 4-block, 23
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
YO-36402-15
More Details
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 6-block, 12
Category: Thermo Scientific Multi-Blok Dry Block Heaters and Blocks
Description: Thermo Scientific Multi-Blok Dry Block Heaters (Analog)
Brand:
Thermo Scientific
Model Number:
400
YO-36402-16
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific Multi-Blok® Analog Dry Block Heater Base; one-bl
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific Multi-Blok Dry Block LED Display Heater Base;, 2-blo (150) (YO-36402-02)
|
Next: Thermo Scientific Multi-Blok® Dry Block LED Display Heater Base; t (2001-1Q)
31125329-p-content_2 31125329pcontent2