Thermo Scientific Nunc CryoFlex ™ Tubing, 15/pack

YO-03755-64
Loading price...

Usually ships in 14 days.
Qty

  • Safely use cryogenic vials in liquid phase of nitrogen
Polyethylene tubing prohibits entrapment of liquified nitrogen inside the vial—reduces risk of possible explosion or biohazard release.

Note: One case (20 packs, each pack has 15 tubing segments) minimum order.
Product Type Cryogenics accessory
Color Clear
Quantity 15/pk
Dimensions 14 mm D x 500 mm L
Brand Thermo Scientific Nunc
Manufacturer number 343958





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for Thermo Scientific Nunc CryoFlex ™ Tubing, 15/pack
    Image
      Product Detail
    SKU

    Thermo Scientific Nunc ™ Thermanox Coverslips, 15 mm diameter; 500/case

    Category: Slides/Cover Slips
    Description: Thermo Scientific Nunc Thermanox Coverslips
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Polycarbonate test tubes; 15-mL; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom tubes; 15-mL; PPCO; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom tubes; 38-mL; PC; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Polycarbonate Round-bottom tubes; 50-mL; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom tubes; 50-mL; PC; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom tubes; 100-mL; PC; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom tubes; 12-mL; PPCO; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom tubes; 38-mL; PPCO; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Thermo Scientific Nunc Round-bottom test tubes; 50-mL; PPCO; 10/pack

    Category: Centrifuge Tubes
    Description: Thermo Scientific Nunc Round-Bottom Tubes
    Brand: Thermo Scientific Nunc


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers

    Do you have questions about this product?

    get answers from real customers and in-house experts with AnswerBox.

    3 Questions | 3 Answers
    Displaying questions 1-3
    • Q:

      If I need to order this, what will be the cost of the product? Is it $89 for just 1 order (all I need is one package) or do I have to pay $1780 for 1 order(i.e. unnecessarily purchasing the whole case of 20 packages)?
      Asked on 5/23/2011 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        We sell this in a pack of 15, 500 mm long, segments. You do not need to purchase the entire case of 20 of these packs.

        Answered on 5/24/2011 by Jen from Technical Support
    • Q:

      heat gun? Please recommend appropriate heat gun for CryoFlex Tubing sealing. Is it possible to use a same heat gun as to use a glue melting. Let me know a heat gun model or picture for this. thanks a lot...
      Asked on 9/16/2010 by joshua from pyeong taek in korea

      1 answer

      • Staff Reviewer

        A:

        CryoFlex tubing is a heat shrink tubing, not a glue type. Please contact directly for further assistance. Contact information may be found under the "contact/about us" tab.

        Answered on 9/20/2010 by Jen from Technical Support
    • Q:

      How do we seal the tube? Is there any equip. needed? I cannot think that the tube is sealed by itself.
      Asked on 9/14/2010 by joshua from Pyeong taek in korea

      1 answer

      • STAFF EXPERT

        A:

        The after the vials are positioned in the CryoFlex tubing, the tubing is gently heated with a heat gun (preferably with a heat-shrink tubing attachment). The heated ends can then be crimped, squeezed or even melted closed. To remove the vial, carefully cut around the cap seal .

        Answered on 9/16/2010 by Josh from Technical Support
    Displaying questions 1-3

    Do you have a question about this product? 

    Product Reviews

    REVIEW SNAPSHOT®

    by PowerReviews
    CryoFlex<tm> Tubing, 15/pack
     
    4.0

    (based on 1 review)

    Reviewed by 1 customer

    Displaying review 1

    Back to top

     
    4.0

    It saved our cells!

    By CVR

    from San Diego, CA

    About Me Academic/Professional Researcher

    Pros

    • Easy To Use

    Cons

    • Tubing bit large for tube

    Best Uses

    • In The Lab

    Comments about CryoFlex<tm> Tubing, 15/pack:

    The tubing was easy to use. The only problem was getting it to fit into our 81-slot freezer boxes when the cryotube was inserted.

    Displaying review 1

    Back to top


    Thermo Scientific Nunc CryoFlex ™ Tubing, 15/pack
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Thermo Scientific Nunc Coverglass F/1838-31 1000/cs  |  Next: Thermo Scientific Nunc Cryotube Cap Inserts 500/bx