Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Temperature recorder 6 circular paper 7 day Celsius scale 115V
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-03816-34
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Accessories
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-03816-30
,
browse similar items
or
Contact us for more information.
Temperature recorder; 6" circular paper; 7-day; Celsius scale; 115V (5016-9 C SCALE)
YO-03816-34
Loading price...
Discontinued
Qty
:
Save to shopping list
Keep a permanent record of temperature on 6" circular chart. A sapphire stylus scribes clear traces on the pressure-sensitive chart paper. Mount recorder in the recessed instrument panel of low-temperature cabinets. Accuracy is as close as 5ºF (2ºC) over the full temperature range. Includes one box of chart paper. Shpg wt 9.5 lb (4.4 kg).
Specifications
Product Type
Accessory
Brand
ScienTemp
Manufacturer number
5016-9 C SCALE
Accessories
Qty
/
Price
Availability
Replacement chart paper, -100 to 0°C; Box of 50
YO-03816-39
BOX of 50
Call for availability.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
LABORATORY EQUIPMENT- Getting Cryogenics Down Cold
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Temperature recorder; 6" circular paper; 7-day; Celsius scale; 115V (5016-9 C SCALE)
Circular Paper
Dickson 3" Compact Temperature Recorders
Temperature Recorders
Portable Temperature Recorder
7 Day Circular Chart Paper
7 Day Recording Thermometer
Thermo Scientific Temperature Recorders
Cole-Parmer Single-Use Temperature Recorders
Madgetech in-Transit Temperature Recorder
High Temperature Recorder
Related Products for Temperature recorder; 6" circular paper; 7-day; Celsius scale; 115V (5016-9 C SCALE)
Image
Product Detail
SKU
Temperature recorder; 6" circular paper; 7-day; Fahrenheit scale; 115V (5016-9 F SCALE)
Category: Accessories
Description: Temperature Recorders
Model Number:
5016-9 F SCALE
YO-03816-24
More Details
Temperature recorder; 6" circular paper; 7-day; Celsius scale; 230V (5016-10 C SCALE)
Category: Accessories
Description: Temperature Recorders
Model Number:
5016-10 C SCALE
YO-03816-35
More Details
Temperature recorder; 6" circular paper; 7-day; Fahrenheit scale; 230V (5016-10 F SCALE)
Category: Accessories
Description: Temperature Recorders
Model Number:
5016-10 F SCALE
YO-03816-25
More Details
3" Compact Temperature Recorder, 24 hour or 7-day, -14 to 32°F (-2
Category: Circular Chart Recorders
Description: Dickson 3" Compact Temperature Recorders
Brand:
Dickson
Model Number:
SC367
YO-80014-00
More Details
3" Compact Temperature Recorder, 24 hr or 7-day, 0 to 50°F (-15 to
Category: Circular Chart Recorders
Description: Dickson 3" Compact Temperature Recorders
Brand:
Dickson
Model Number:
SC377
YO-80014-02
More Details
3" Compact Temperature Recorder, 24 hour or 7-day, 22 to 68F (-5 to 20
Category: Circular Chart Recorders
Description: Dickson 3" Compact Temperature Recorders
Brand:
Dickson
Model Number:
SC386
YO-80014-04
More Details
3" Compact Temperature Recorder, 24 hr or 7-day, 50 to 96°F (10 to
Category: Circular Chart Recorders
Description: Dickson 3" Compact Temperature Recorders
Brand:
Dickson
Model Number:
SC387
YO-80014-06
More Details
3" Compact Temperature Recorder, 24 hour or 7-day, 76 to 122°F (25
Category: Circular Chart Recorders
Description: Dickson 3" Compact Temperature Recorders
Brand:
Dickson
Model Number:
SC397
YO-80014-08
More Details
6" Circular Temperature/Humidity Recorder, 115 VAC
Category: 6 inch Multifunctional Two-Channel Recorder
Description: Circular Temperature/Humidity Recorder
Brand:
SUPCO
Model Number:
CR4 + KTH
YO-80055-00
More Details
6" Circular Temperature/Humidity Recorder, 220 VAC
Category: 6 inch Multifunctional Two-Channel Recorder
Description: Circular Temperature/Humidity Recorder
Brand:
SUPCO
Model Number:
CR4220 + KTH
YO-80055-05
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Temperature recorder; 6" circular paper; 7-day; Celsius scale; 115V (5016-9 C SCALE)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Temperature Range -20/70c; in-transit single use temp rec
|
Next: Temperature recorder; 6" circular paper; 7-day; Celsius scale; 230V (5016-10 C SCALE)
31125329-p-content_2 31125329pcontent2