Connection Tube For Ln2 Tank (05.871.0001)

YO-04182-26
$3,315.45 AUD/EACH

Call for availability.
Qty

  • Fast, efficient cryogenic grinding at -196°C—ideal for plastics!
Product Type Accessory
Brand Retsch
Manufacturer number 05.871.0001





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for Connection Tube For Ln2 Tank (05.871.0001)
    Image
      Product Detail
    SKU

    Push-to-connect stem-to-hose adapters, 1/4" x 5/16", 10/pack (YO-34007-05)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 1/4" x 1/4", 10/pack (YO-34006-79)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 3/8" x 1/4", 10/pack (YO-34006-80)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 3/8" x 3/8", 10/pack (YO-34006-81)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 1/2" x 1/2", 10/pack (YO-34006-83)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 1/4" x 3/16", 10/pack (YO-34007-04)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 5/16" x 3/16", 10/pack (YO-34007-06)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 5/16" x 1/4", 10/pack (YO-34007-07)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 5/16" x 3/8", 10/pack (YO-34007-08)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Push-to-connect stem-to-hose adapters, 3/8" x 5/16", 10/pack (YO-34007-09)

    Category: Swivel adapters for John Guest acetal push-to-connect fittings
    Description: John Guest Acetal push-to-connect tube-to-hose barb adapters
    Brand: John Guest


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews