Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-04560-24
Heavy-duty dual clamp
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-04560-24
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Accessories for Laboratory Mixers
Share
|
Heavy-duty dual clamp (YO-04560-24)
YO-04560-24
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Clamps ensure sturdy connection between mixers and stands.
Specifications
Product Type
Accessory
Material
Stainless Steel
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for Heavy-duty dual clamp (YO-04560-24)
Heavy Duty Hose Clamps
Troemner Heavy-Duty Tapered Clamps
Heavy Duty C Clamp
Heavy Duty Lab Clamps
Heavy Duty Magnetic Stirrers
Troemner Heavy-Duty Three-Prong Utility Clamps
Cole-Parmer Drum/Barrel Heavy-Duty Mixers
Cole-Parmer Heavy-Duty
Oakton Heavy-Duty Extra-Long Penetration Probes
Heavy Duty Carts
Related Products for Heavy-duty dual clamp (YO-04560-24)
Image
Product Detail
SKU
Heavy Duty Tapered Clamp, Small (915290)
Category: Specialty Clamps and Supports
Description: Heavy-Duty Tapered Clamps
Brand:
Troemner
Model Number:
915290
YO-08047-60
More Details
Ohaus Heavy-Duty Dual-Beam Mechanical Balance, 20 kg x 1 g
Category: Mechanical Balances
Description: Ohaus Heavy-Duty Dual-Beam Mechanical Balance
Brand:
Ohaus
Model Number:
1119-D0
YO-01042-01
More Details
Heavy Duty Tapered Clamp, Large (915291)
Category: Specialty Clamps and Supports
Description: Heavy-Duty Tapered Clamps
Brand:
Troemner
Model Number:
915291
YO-08047-65
More Details
Heavy-Duty Bath, HDPE, 7 gal
Category: Trays
Description: Heavy-Duty Baths, HDPE
Brand:
Cole-Parmer
Model Number:
P339-01-2510
YO-06712-10
More Details
Heavy-Duty Bath, HDPE, 15 gal
Category: Trays
Description: Heavy-Duty Baths, HDPE
Brand:
Cole-Parmer
Model Number:
P337-01-2510
YO-06712-30
More Details
Heavy-Duty Smart™ Container, HDPE, 30 gal
Category: Mobile Bins and Tubs
Description: Heavy-Duty Smart Container, HDPE
Brand:
Cole-Parmer
Model Number:
P300W/C
YO-06740-50
More Details
Heavy duty, gear-drive mixer; 1/2-hp, 115/230 VAC, 36"L shaft, 8" dual (YO-50311-24)
Category: Heavy-Duty Gear-Drive Batch Mixers
Description: Heavy-Duty, Gear-Drive Electric (TEFC) Powered Mixers
Brand:
Cole-Parmer
YO-50311-24
More Details
Heavy-duty clamp (YO-04340-50)
Category: Laboratory Frame/Rod/Clamp Holders
Description: Heavy-Duty Holders
Brand:
Troemner
YO-04340-50
More Details
Heavy-Duty Three-Prong extension clamp, Dual, Small
Category: Utility Clamps
Description: Heavy-Duty Three-Prong Utility Clamps
Brand:
Troemner
Model Number:
916113
YO-08021-12
More Details
Heavy-Duty Three-Prong extension clamp, Dual, Medium
Category: Utility Clamps
Description: Heavy-Duty Three-Prong Utility Clamps
Brand:
Troemner
YO-08021-14
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Heavy-duty dual clamp (YO-04560-24)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Heavy-duty DO probe, 25-meter cable
|
Next: Heavy-duty Lab Frame, Aluminum
31125329-p-content_2 31125329pcontent2