Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific Cimarec Telemodul 40C 40 Watt Control Unit 100 240V
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-04651-05
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Magnetic Stirrers
Share
|
Thermo Scientific Cimarec Telemodul 40C, 40 Watt Control Unit 100-240V
YO-04651-05
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Compatible with Cimarec stirrers
10 power settings
RS-232 interface for data transfer and PC control
TC-IP network integration option
7-Watt controller (04651-01, -03) is a compact controller ideal for small to medium volumes of low-viscosity liquids. It features continuous speed selection and automatic start-up for safe stirrer acceleration.
20-Watt controller (04651-04) is a menu-driven microprocessor controlled unit. The knob is easily controlled with one hand by pushing down and rotating. Recent settings are stored in the memory. These controllers feature three progam keys for individually programming rapid start up and a shake mode for vertical mixing.
40-Watt Controller (04651-05) offers all the same functions as the 20-Watt controller with a higher power output of 40 Watts for more demanding applications. This model has ten power settings and includes RS-232 interface for data transfer and PC control. Additionally, it has a TC-IP network integration option.
The Telemodul 40 C controller offers all the functionality of the Telemodul 20 C, with power output boosted to 40 Watts for more demanding stirring tasks.
What's Included:
6-ft power cord.
Specifications
Product Type
Stirrer Accessories
Speed (rpm) (low)
100
Speed (rpm) (high)
2000
Dimensions
6 in x 3.75 in x 6.5 in
Max stirring volume (Liters)
0
Plug type
U.S. std
Model
50090774
CE Compliance
Yes
Brand
Thermo Scientific Cimarec
Manufacturer number
50090774
Power
100-240 VAC, 50/60Hz, .2 Amps, 40 Watts
Technical Resources
Related technical articles (Roll over for summary)
Stirrers and Hot Plates Selection Guide
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Thermo Scientific Cimarec Telemodul 40C, 40 Watt Control Unit 100-240V
Thermo Scientific Cimarec Compact Stirrers
Thermo Scientific Cimarec Multiposition Electromagnetic Stirrers
Thermo Scientific Cimarec Multiposition Stirrers
Thermo Scientific Cimarec Large Capacity Stirrers
Thermo Scientific Cimarec Immersible Stirrers
Thermo Scientific Cimarec Electromagnetic Stirrers
Thermo Scientific Cimarec Stirring Hot Plates
Thermo Scientific Cimarec Digital Hot Plate
Thermo Scientific Cimarec Single
Thermo Scientific Cimarec External Remote Controllers
Related Products for Thermo Scientific Cimarec Telemodul 40C, 40 Watt Control Unit 100-240V
Image
Product Detail
SKU
Thermo Scientific Cimarec Mini with 7-watt controller 115V
Category: Compact Stirrers
Description: Thermo Scientific Cimarec Mini and Micro Stirrers
Brand:
Thermo Scientific Cimarec
Model Number:
50088118
YO-04651-44
More Details
Thermo Scientific Cimarec Micro, with 7-watt controller, 115 V
Category: Compact Stirrers
Description: Thermo Scientific Cimarec Mini and Micro Stirrers
Brand:
Thermo Scientific Cimarec
Model Number:
50088150
YO-04651-56
More Details
Thermo Scientific Cimarec Telemodul 20 C, 20 Watt Control Unit 100-240
Category: Magnetic Stirrers
Description: Thermo Scientific Cimarec External Remote Controllers
Brand:
Thermo Scientific Cimarec
Model Number:
50090773
YO-04651-04
More Details
Thermo Scientific Cimarec Telemodul 40M Controllers 230V Germany
Category: Analog Stirrers
Description: Thermo Scientific Cimarec Stirrer Controllers
Brand:
Thermo Scientific
Model Number:
50118901
YO-04651-08
More Details
Thermo Scientific Cimarec Telemodul 40M Controllers 115V US
Category: Analog Stirrers
Description: Thermo Scientific Cimarec Stirrer Controllers
Brand:
Thermo Scientific
Model Number:
50119123
YO-04651-09
More Details
Thermo Scientific Cimarec Telemodul 80M Controllers 230V Germany
Category: Analog Stirrers
Description: Thermo Scientific Cimarec Stirrer Controllers
Brand:
Thermo Scientific
Model Number:
50118908
YO-04651-11
More Details
Thermo Scientific Cimarec Telemodul 80M Controllers 115V US
Category: Analog Stirrers
Description: Thermo Scientific Cimarec Stirrer Controllers
Brand:
Thermo Scientific
Model Number:
50119125
YO-04651-12
More Details
Thermo Scientific Cimarec i Telemodul controller 230V EU
Category: Magnetic Stirrers
Description: Thermo Scientific Cimarec External Remote Controllers
Brand:
Thermo Scientific Cimarec
Model Number:
50119115
YO-04651-01
More Details
Thermo Scientific Cimarec i Telemodul controller 115 US
Category: Magnetic Stirrers
Description: Thermo Scientific Cimarec External Remote Controllers
Brand:
Thermo Scientific Cimarec
Model Number:
50119119
YO-04651-03
More Details
Thermo Scientific Cimarec Maxi 40, with Telemodul 40 C, 40 Watt Contro
Category: Compact Stirrers
Description: Thermo Scientific Cimarec Compact and Maxi Stirrers
Brand:
Thermo Scientific Cimarec
Model Number:
50088122
YO-04651-76
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific Cimarec Telemodul 40C, 40 Watt Control Unit 100-240V
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific Cimarec Telemodul 20 C, 20 Watt Control Unit 100-240
|
Next: Thermo Scientific Cimarec Telemodul 40M Controllers 115V US
31125329-p-content_2 31125329pcontent2