Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific Cimarec ceramic stirring hot plate 7
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-04660-85
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17002-27
Innocal NIST Calibration for Stirrers and Heat
$297.84 AUD/EACH/EACH
Usually ships in 30 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Stirring Hot Plates
Share
|
Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131320-33Q)
YO-04660-85
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Microprocessor control provides constant speed
StirTrac
braking feature brings stir bar to an immediate stop for quick flask removal
StirTrac
technology offers improved slow speed stirring, consistent speed control, and stronger magnetic coupling. Durable low-profile, cast aluminum hot plate stirrers are complete with a seamless reflective white ceramic top that provides visibility, an alkali/acid reisistance surface, and is easy to clean. Intergral ring-stand holder accomodates 0.5" diameter support rod. Digital hot plate stirrer features a LED that indicates temperature setting in 5° increments. Patented warning light signal system goes off when heating surface is 50°C(122°F); protects user from accidental burns. Detachable cord is included.
Specifications
Product Type
Stirrer (Hot Plates)
Max load capacity (lbs)
25
Speed (rpm) (low)
50
Speed (rpm) (high)
1200
Max temperature (º C)
540
Top plate dimensions (in.)
7x7
Dimensions
8.25 in W x 3.75 in H x 13 in D
Max stirring volume (Liters)
20
Plug type
European
Model
SP131320-33Q
CE Compliance
Yes
Brand
Thermo Scientific Cimarec
Manufacturer number
SP131320-33Q
Power
230 VAC, 50 Hz, 4.9 Amps, 1170 Watts
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Stirrers and Heat
YO-17002-27
EACH
Usually ships in 30 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131320-33Q)
Thermo Scientific Super-Nuova Stirring Hot Plates
Thermo Scientific Thermolyne Cast-Aluminum Hot Plates
Thermo Scientific Cimarec Ceramic 10x10 Stirrer
Ceramic Stirring Hot Plate
Thermo Scientific Cimarec Stirring Hot Plates
Thermo Scientific Cimarec Hot Plates
Thermo Scientific Digital Stirring Hot Plate
Thermo Scientific Rt Stirring Hot Plate
Thermo Scientific Ceramic Top Hot Plates
Thermo Scientific Glass Ceramic Hot Plate
Related Products for Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131320-33Q)
Image
Product Detail
SKU
Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131325Q)
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP131325Q
YO-04660-80
More Details
Thermo Scientific Cimarec aluminum stirring hot plate, 7 (SP142020-33Q)
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP142020-33Q
YO-04660-87
More Details
Thermo Scientific Cimarec ceramic stirring hot plate, 4x4 120v
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP131015Q
YO-04660-70
More Details
Thermo Scientific Cimarec ceramic stirring hot plate, 4x4 240v
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP131010-33Q
YO-04660-75
More Details
Thermo Scientific Cimarec aluminum stirring hot plate, 7 (SP142025Q)
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP142025Q
YO-04660-82
More Details
Thermo Scientific Cimarec ceramic stirring hot plate, 10x10 120v
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP131635Q
YO-04660-90
More Details
Thermo Scientific Cimarec ceramic stirring hot plate, 10x10 240v
Category: Stirring Hot Plates
Description: Thermo Scientific Cimarec Stirring Hot Plates
Brand:
Thermo Scientific Cimarec
Model Number:
SP131630-33Q
YO-04660-95
More Details
Thermo Scientific Super-Nuova™ ceramic stirring hot plate, 7 x 7 12
Category: Stirring Hot Plates
Description: Thermo Scientific Super-Nuova Stirring Hot Plates
Brand:
Thermo Scientific Super-Nuova
Model Number:
SP131825Q
YO-86576-50
More Details
Thermo Scientific Super-Nuova™ ceramic stirring hot plate, 7 x 7 22
Category: Stirring Hot Plates
Description: Thermo Scientific Super-Nuova Stirring Hot Plates
Brand:
Thermo Scientific Super-Nuova
Model Number:
SP131820-33Q
YO-86576-55
More Details
Thermo Scientific Super-Nuova™ ceramic stirring hot plate, 10 x 10 (SP133835Q)
Category: Stirring Hot Plates
Description: Thermo Scientific Super-Nuova Stirring Hot Plates
Brand:
Thermo Scientific Super-Nuova
Model Number:
SP133835Q
YO-86576-70
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131320-33Q)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific Cimarec ceramic stirring hot plate, 4x4 240v
|
Next: Thermo Scientific Cimarec ceramic stirring hot plate, 7 (SP131325Q)
31125329-p-content_2 31125329pcontent2