Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM (YO-04680-00)

YO-04680-00
Loading price...

Usually ships in 14 days or sooner.
Qty
    Safely use these air-drive mixers whenever electrical power is hazardous or impractical. Ideal for mixing solvents, lacquers, and volatile chemicals without the possibility of spark ignition.
    Mixers accept an air input line pressure to 80 or 100 psi (10 to 12 cfm). A needle valve enables you to control mixing speed throughout the range for each mixer, and a built-in muffler ensures quiet operation. We recommend installing an air filter/regulator 07042-75 and an air lubricator 07042-70 in your air supply line.Includes: 316 SS paddle assembly (3⁄8" dia x 12"L [10 mm dia x 305 mm L] shaft with 21⁄2" [64 mm] dia propeller), collet, 10"L x 9⁄16" dia (254 mm L x 14 mm dia) mounting rod, muffler, needle valve, and 4-ft (1.2-m) L air hose with fittings.
    Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
    Product Type Mixer
    Min speed (rpm) 200
    Max speed (rpm) 10000
    Motor (hp) 0.33
    Max torque (in-oz) 64

    Accessories
    Qty/Price
    Availability

    EACH

    Usually ships in 14 days or sooner.

    EACH

    Call for availability.
    Calibrations
    Qty/Price
    Availability

    EACH
    Usually ships in 18 days.




    Technical Resources
     
    Related Products for Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM (YO-04680-00)
    Image
      Product Detail
    SKU

    Low-cost, Direct-drive Air Motor Mixer; 200-7,000 RPM (YO-04686-00)

    Category: Hazardous-Duty Air-Drive Mixers
    Description: Hazardous-Duty Direct-Drive Air Motor Mixers
    Brand: Guaranteed In Stock Program


    Low-cost, Direct-drive Air Motor Mixer; 300-3,000 RPM (YO-04686-15)

    Category: Hazardous-Duty Air-Drive Mixers
    Description: Hazardous-Duty Direct-Drive Air Motor Mixers
    Brand: Cole-Parmer


    Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)

    Category: Hazardous-Duty Air-Drive Mixers
    Description: Hazardous-Duty Gear-Drive Air Motor Mixers
    Brand: Cole-Parmer


    Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)

    Category: Hazardous-Duty Air-Drive Mixers
    Description: Hazardous-Duty Gear-Drive Air Motor Mixers
    Brand: Cole-Parmer


    Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00)

    Category: Hazardous-Duty Air-Drive Mixers
    Description: Hazardous-Duty Gear-Drive Air Motor Mixers
    Brand: Guaranteed In Stock Program


    Low-cost, gear-drive air motor mixer; 50-1000 rpm (YO-04686-10)

    Category: Hazardous-Duty Air-Drive Mixers
    Description: Hazardous-Duty Gear-Drive Air Motor Mixers
    Brand: Cole-Parmer


    Low-Cost Analog Incubator, 0.7 cubic ft, Steel Door, 115 VAC (YO-03613-00)

    Category: General Purpose Incubators
    Description: Low-Cost Incubators
    Brand: Cole-Parmer


    Lightnin direct-drive pressure batch mixers, Air (YO-50310-68)

    Category: Lightnin Batch Industrial Mixers
    Description: Lightnin Direct-Drive Pressure Batch Industrial Mixers
    Brand: Lightnin


    Lightnin direct-drive pressure batch mixers, Air (YO-50323-50)

    Category: Lightnin Batch Industrial Mixers
    Description: Lightnin Direct-Drive Pressure Batch Industrial Mixers
    Brand: Lightnin


    Lightnin direct-drive pressure batch mixer, TENV (YO-50310-70)

    Category: Lightnin Batch Industrial Mixers
    Description: Lightnin Direct-Drive Pressure Batch Industrial Mixers
    Brand: Lightnin


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers

    Do you have questions about this product?

    get answers from real customers and in-house experts with AnswerBox.

    4 Questions | 4 Answers
    Displaying questions 1-4
    • Q:

      The answer to the question concerning the dimensions of this mixer are not clear to me:
      The mixer itself is about 4.235" tall and has a diameter of 2.44"

      Is it possible to publish the dimensions in metres / centimetres ?
      Asked on 7/27/2012 by kat from NL

      1 answer

      • STAFF EXPERT

        A:

        Our technical database gives the dimensions in height/length/width as 6.53" x 12.26" x 9.93". Converted to centimeters that is 16.59cm x 31.14cm x 25.22cm. These dimensions include the impeller, shaft, mounting rod and motor.

        Answered on 7/27/2012 by Jon from Technical Support
    • Q:

      Are there longer shaft lengths available for this mixer? 24"
      Asked on 6/19/2012 by Anonymous

      1 answer

      • STAFF EXPERT

        A:

        Item 04552-00, which is 24 inches long, and Item 4552-05, at 30 inches long are sold separately.

        Answered on 6/19/2012 by Rita from Customer Service
    • Q:

      where do I put oil in this particular mixer
      Asked on 9/2/2011 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        Detergent SAE #10 automotive oil (or Gast AD220) should be used to lubricate all moving parts, but this is not an engine that requires oil or has an oil reservoir. This is a motor that utilizes compressed air to move the motor.
        Please call for further assistance.

        Answered on 9/6/2011 by Jen from Technical Support
    • Q:

      what are the physical dimensions of the mixing motor not including the mixing shaft?

      Is there some sort of mounting plate?
      Asked on 6/16/2010 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        There is a mounting rod used for this mixer. The mounting rod is 10" long and 9/16" in diameter.
        The mixer itself is about 4.235" tall and has a diameter of 2.44"

        Answered on 6/18/2010 by Jen from Technical Support
    Displaying questions 1-4

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought

    Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM (YO-04680-00)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Low-Cost Waterproof T-Shaped Pocket Thermometer (RT610B)  |  Next: Low-cost, Direct-drive Air Motor Mixer; 200-7,000 RPM (YO-04686-00)