Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips 1 2 dia
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-04711-83
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Accessories for Sonicator ® Ultrasonic Processors
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-04712-66
,
browse similar items
or
Contact us for more information.
Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1/2"
YO-04711-83
Loading price...
Discontinued
Qty
:
Save to shopping list
Probe selection is determined by sample volume, sample type, and the desired intensity. As tip diameter increases, the intensity/amplitude decreases. Tips erode over time and replacement tips are available separately.
Specifications
Product Type
Ultrasonic Processor Probes
Tip Diameter (in.)
1/2
Max sample size (mL)
250
Brand
Qsonica
Accessories
Qty
/
Price
Availability
Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia
YO-04711-86
EACH
Call for availability.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Ultrasonic Homogenizer Selection Guide
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1/2"
Sonicator Replacement Probes with Threaded Tips
Ultrasonic Cell Disruptor
Potted Threaded Insert 1 2
Replacement Probes
Cole-Parmer Replacement Threaded Tips
Replacement Threaded Tips
Replacement Electrode Phtestr 1 2
Medical Temperature Probe W Ecg Tips
Replacement Probe for Ph Meter
Thermometers Replacement Probe
Related Products for Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1/2"
Image
Product Detail
SKU
Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1" di
Category: Accessories for Sonicator
®
Ultrasonic Processors
Description: Sonicator Replacement Probes with Threaded Tips
Brand:
Qsonica
Model Number:
210
YO-04711-85
More Details
Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 3/4"
Category: Accessories for Sonicator
®
Ultrasonic Processors
Description: Sonicator Replacement Probes with Threaded Tips
Brand:
Qsonica
Model Number:
207
YO-04711-84
More Details
Ultrasonic Cell Disruptor Replacement Tips, 1/2" dia
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonicator Replacement Probes with Threaded Tips
Brand:
Qsonica
Model Number:
406
YO-04711-86
More Details
Ultrasonic Cell Disruptor Replacement Tips, 3/4" dia
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonicator Replacement Probes with Threaded Tips
Brand:
Qsonica
Model Number:
407
YO-04711-87
More Details
Ultrasonic Cell Disruptor Replacement Tips, 1" dia
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonicator Replacement Probes with Threaded Tips
Brand:
Qsonica
Model Number:
408
YO-04711-88
More Details
Replacement Threaded Tip 1/2" (flat) (YO-04710-40)
Category: Accessories for Cole-Parmer 500-, and 750-Watt Processors
Description: Replacement Threaded Tips
Brand:
Cole-Parmer
YO-04710-40
More Details
Ultrasonic Cell Disruptor Microtip Probes, 1-16" dia
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonicator Microtip Probes
Brand:
Qsonica
Model Number:
418
YO-04711-89
More Details
Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonicator Microtip Probes
Brand:
Qsonica
Model Number:
419B
YO-04711-92
More Details
Qsonica Ultrasonic Cell Disruptor Cup Horn Stand
Category: Accessories for Sonicator Ultrasonic Processors
Description: Qsonica Sonicator Ultrasonic Cell Disruptor Cup Horn Stand
Brand:
Qsonica
Model Number:
448
YO-04711-97
More Details
Ultrasonic Cell Disruptor Microtip Stepped Probe Replacement Tip for 0
Category: Accessories for Sonicator Ultrasonic Processors
Description: Sonicator Microtip Probes
Brand:
Qsonica
Model Number:
422
YO-04711-94
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1/2"
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 1" di
|
Next: Ultrasonic Cell Disruptor Replacement Probes with Threaded Tips, 3/4"
31125329-p-content_2 31125329pcontent2