Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Qsonica Touch Screen Sonicator Ultrasonic Processor 220V
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-04712-81
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Misonix Ultrasonic Processors
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-04712-84
,
browse similar items
or
Contact us for more information.
Misonix Sonicator 4000 Ultrasonic Processor (S-4000-010)
YO-04712-81
Loading price...
Discontinued
Qty
:
Save to shopping list
Run multiple programs in sequence
User-friendly touch screen control
Wide variety of applicationsdisrupt cells, shear DNA, mix compounds, extract samples, and nanoparticle dispersion
Self diagnostics and help screens simplify operation
The Sonicator tracks frequency changes in the convertor/tip assembly caused by load and temperature changes, and maintains optimal electrical efficiency at all times. Updated features include improved frequency and amplitude control, multiple sequence programming, and the ability to more accurately track the total energy delivered to your sample. The digital design allows the system to run more efficiently and requires less power to effectively process samples. This reduced energy consumption allows the unit to run cooler which extends the life of internal components. Pulse on/off from 1 second to 24 hours and total run time up to 72 hours.
The ½” probe which comes with the 04712-79 and 04712-81 is appropriate for volumes of 10-250 mL. The processor itself, with the appropriate accessories, can do volumes within other ranges. For example, smaller sample volumes down to 0.1 mL can be processed with the 04711-95 & 04711-96 cup horn; larger sample volumes can be processed at a rate of 40 L/hr with the 04711-98 continuous flow cell.
What's Included:
convertor with cable, ½” horn, wrench set, and 6-ft power cable. Order additional probes and accessories separately below.
Specifications
Product Type
Ultrasonic Processor
Description
Qsonica Touch-Screen Sonicator Ultrasonic Processor, 220V
Max sample size (mL)
40,000
Watts
600
Power (VAC)
220
Dimensions (" W)
8.5
Dimensions (" H)
10.5
Dimensions (" D)
19
Manufacturer number
S-4000-010
Model
S-4000-010
Brand
Qsonica
Technical Resources
Related technical articles (Roll over for summary)
Ultrasonic Homogenizer Selection Guide
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Misonix Sonicator 4000 Ultrasonic Processor (S-4000-010)
Misonix Sonicator S-4000 Ultrasonic Processors
Sonicator Qsonica Ultrasonic Processors
Cole-Parmer Accessories for 130-Watt Ultrasonic Processors
Ultrasonic Processor Sound Enclosures
Ultrasonic Processor Stand and Converter Clamps
Cole-Parmer Ultrasonic Processor Sound-Abating Enclosure
Ultrasonic Processor Booster
Qsonica Q55 Sonicator Ultrasonic Processor
Sonicator Ultrasonic Processors
Qsonica Accessories for Sonicator Ultrasonic Processors
Related Products for Misonix Sonicator 4000 Ultrasonic Processor (S-4000-010)
Image
Product Detail
SKU
Misonix Sonicator 4000 Ultrasonic Processor (S-4000)
Category: Misonix Ultrasonic Processors
Description: Misonix Sonicator S-4000 Ultrasonic Processors
Brand:
Sonicator
Model Number:
S-4000
YO-04712-79
More Details
Ultrasonic Processor Sound-Abating enclosure; 30"H
Category: Accessories for Cole-Parmer 500-, and 750-Watt Processors
Description: Ultrasonic Processor Sound-Abating Enclosure
Brand:
Cole-Parmer
YO-04710-48
More Details
Eight-element probe for 130-Watt Cole-Parmer ultrasonic processors (630-0602)
Category: Accessories for Cole-Parmer 130-Watt Ultrasonic Processors
Description: Eight-Element Probe for Cole-Parmer 130-Watt Ultrasonic Processors
Brand:
Cole-Parmer
Model Number:
630-0602
YO-04712-08
More Details
Ultrasonic Processor Sound-Abating Enclosure, 20"H
Category: Accessories for Cole-Parmer 130-Watt Ultrasonic Processors
Description: Ultrasonic Processor Sound Enclosure
Brand:
Cole-Parmer
Model Number:
630-0451
YO-04712-50
More Details
Open end wrench (requires 2) for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-60
More Details
Coupler (for Probe 04712-65) for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-64
More Details
Cup horn (3") for use with ultrasonic processors
Category: Qsonica Ultrasonic Processors
Description: Qsonica Cup Horns for Touch-Screen Sonicator
Brand:
Sonicator
YO-04712-75
More Details
Cup horn (5.5") for use with ultrasonic processors
Category: Qsonica Ultrasonic Processors
Description: Qsonica Cup Horns for Touch-Screen Sonicator
Brand:
Sonicator
YO-04712-76
More Details
Replacement tube holder (8 tubes) for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-77
More Details
Replacement tube holder (20 tubes) for use with ultrasonic processors
Category: Qsonica Q55 Sonicator ultrasonic processor
Description: Accessories for the Qsonica Q55 Sonicator ultrasonic processor
Brand:
Sonicator
YO-04712-78
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Misonix Sonicator 4000 Ultrasonic Processor (S-4000-010)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Minotome PLUS™ Cryostat, 200-240 VAC, 50Hz
|
Next: Misonix Sonicator 4000 Ultrasonic Processor (S-4000)
31125329-p-content_2 31125329pcontent2