Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
YSI BOD probe adapter
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-05510-62
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
YSI Meter Accessories
Share
|
YSI BOD probe adapter (5011)
YO-05510-62
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
This probe adapter works with probes 05521-00 and 05519-42.
Note: The adapter is required when using probe 05519-42 with YSI 5100 meters (05510-22,-27).
Specifications
Model
5011
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for YSI BOD probe adapter (5011)
Ysi Bod Meter and Probe
Ysi Multiparameter Probe
Ysi Oxygen Probe
Probe Adapters
Ysi Ph Probe Issues
Ysi Ph Probe
Ysi Proobod Probe
Ph Probe Adapter
Ysi Temperature Probes
Ysi Conductivity Probes
Related Products for YSI BOD probe adapter (5011)
Image
Product Detail
SKU
Self-stirring BOD probe, direct connect to YSI 5000/5100 meters (5010)
Category: Dissolved Oxygen Probes and Sensors
Description: YSI
®
BOD probes
Brand:
YSI
Model Number:
5010
YO-05510-60
More Details
YSI ProOBOD probe, US version
Category: Dissolved Oxygen (DO) Probes and Sensors
Description: YSI ProOBOD probe
Brand:
YSI
YO-59353-30
More Details
YSI ProOBOD probe, international version
Category: Dissolved Oxygen (DO) Probes and Sensors
Description: YSI ProOBOD probe
Brand:
YSI
YO-59353-31
More Details
RTD Probe Adapter; pins are 0.31" apart (93400-50)
Category: RTD Probes and Accessories
Description: Probe Adapters, Connectors, and Extension Cables
Brand:
Cole-Parmer
Model Number:
93400-50
YO-93400-50
More Details
RTD Probe Adapter; pins are 0.44" apart (93400-52)
Category: RTD Probes and Accessories
Description: Probe Adapters, Connectors, and Extension Cables
Brand:
Cole-Parmer
Model Number:
93400-52
YO-93400-52
More Details
Nonstirring BOD bottle probe for YSI® meters (5750)
Category: Dissolved Oxygen Probes and Sensors
Description: YSI
®
BOD probes
Brand:
YSI
Model Number:
5750
YO-05519-10
More Details
Self-stirring BOD bottle probe, 115 VAC, for YSI® meters (5905-115)
Category: Dissolved Oxygen Probes and Sensors
Description: YSI
®
BOD probes
Brand:
YSI
Model Number:
5905-115
YO-05519-42
More Details
YSI IDS Pro OBOD (optical BOD) probe
Category: Benchtop Multiparameter meters
Description: YSI MultiLab IDS Sensors and Cables
Brand:
YSI
YO-05510-54
More Details
Adapter, ANSI 3-pin male miniconnector to Cole-Parmer 3-pin male (011900M)
Category: RTD Probes and Accessories
Description: Probe Adapters, Connectors, and Extension Cables
Brand:
Cole-Parmer
Model Number:
011900M
YO-93400-54
More Details
Cole-Parmer LabGEN Tip Probes Adapter
Category: Homogenizer Probes, and Accessories
Description: Cole-Parmer LabGEN Homogenizer Accessories
Brand:
Cole-Parmer
YO-04727-88
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
YSI BOD probe adapter (5011)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: YSI 900 Free and Total Chlorine Colorimeter Kit
|
Next: YSI COD Colorimeter Kit
31125329-p-content_2 31125329pcontent2