Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Replacement YSI DO membrane cap kit 1 00 mil PTFE
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-05519-61
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
YSI Meter Accessories
Share
|
Replacement YSI DO membrane cap kit, 1.00 mil PTFE (5906)
YO-05519-61
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
The membrane cap kit works with YSI model 58, 52, 5000, and 5100 meters. This kit includes 6 membranes, sanding disc, and 5.25 g of KCl crystals.
Specifications
Model
5906
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for Replacement YSI DO membrane cap kit, 1.00 mil PTFE (5906)
Ysi do Membrane Cap Probe
Replacement Membrane Caps
Membrane Mwco (Daltons) 100
Replacement Ampoules for Colorimetric Test Kits
100 Ml Ptfe Burette
Ysi Replacement Membrane Kits
Ysi Replacements Accessories
Ptfe Replacements Accessories
Probe Cap
Replacement Vials
Related Products for Replacement YSI DO membrane cap kit, 1.00 mil PTFE (5906)
Image
Product Detail
SKU
Spectra/Por Dialysis Membrane Trial Kit, 6000-8000Dalton 32mm
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Standard RC Dialysis Trial Kits
Brand:
Spectra/Por
YO-02900-00
More Details
Spectra/Por Dialysis Membrane Trial Kit, 6000-8000Dalton 50mm
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Standard RC Dialysis Trial Kits
Brand:
Spectra/Por
YO-02900-01
More Details
Spectra/Por Dialysis Membrane Trial Kit, 10,000-12,000Dalton 25mm
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Standard RC Dialysis Trial Kits
Brand:
Spectra/Por
YO-02900-02
More Details
Spectra/Por Dialysis Membrane Trial Kit, 10,000-12,000Dalton 50mm
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Standard RC Dialysis Trial Kits
Brand:
Spectra/Por
YO-02900-03
More Details
Spectra/Por Dialysis Membrane Trial Kit, 3,500 Dalton 18mm
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Standard RC Dialysis Trial Kits
Brand:
Spectra/Por
YO-02900-04
More Details
Spectra/Por Dialysis Membrane Trial Kit, 3,500 Dalton 54mm
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Flat Discs and Sheets
Description: Standard RC Dialysis Trial Kits
Brand:
Spectra/Por
YO-02900-05
More Details
YSI BOD probe adapter (5011)
Category: YSI Meter Accessories
Description: YSI DO Membrane Cap Probe
Brand:
YSI
Model Number:
5011
YO-05510-62
More Details
Membrane Service Kit with 2-mil thick PTFE Membranes (YO-05643-51)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
PTFE
YO-05643-51
More Details
Cole-Parmer Spectroscopy Standard Kit 1, includes 4 pcs, for UV-VIS Ph
Category: Calibration Products and Standards
Description: Cole-Parmer Spectroscopy Standards Kits for UV-Visible Spectrophotometers
Brand:
Cole-Parmer
Model Number:
KIT 1
YO-02660-01
More Details
Spectra/Por 1 Membranes; flat width, 23mm; diameter, 14.6mm; length, 1 (YO-02900-10)
Category: Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
Description: Spectra/Por 1 Regenerated Cellulose (RC) Dialysis Tubing
Brand:
Spectra/Por
YO-02900-10
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Replacement YSI DO membrane cap kit, 1.00 mil PTFE (5906)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Replacement windscreen
|
Next: Replacement/extension dipper shaft (YO-06282-19)
31125329-p-content_2 31125329pcontent2