Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-05521-00
Field DO probe for YSI
®
meters
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
MSDS
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-05521-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
YSI Meter Accessories
Share
|
Field DO probe for YSI® meters (5739)
YO-05521-00
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Dissolved Oxygen Field Probe is both temperature and pressure compensated. Probe includes standard membrane kit 05522-00. Customize the probe with the cable length for your application.
Specifications
Model
5739
Brand
YSI
Accessories
Qty
/
Price
Availability
Standard DO membrane kit for DO probe 05521-00, pack of 30 membranes and 5.25 g of KCl crystals
YO-05522-00
EACH
Call for availability.
Add to Cart
Technical Resources
MSDS 1 (135 B)
Related Searches for Field DO probe for YSI® meters (5739)
Ysi Meters
Ysi Meter Accessories
Ysi Meter Water Quality Package
Replacement Probe for Ph Meter
Temperature Probe Corning Ph Meter False
Probe Carbon Monoxide Meter
Replacement do Probe for Sension Meter
Temperature Probe for Extech Meter
Field Meter
Field Oxygen Meter
Related Products for Field DO probe for YSI® meters (5739)
Image
Product Detail
SKU
Nonstirring BOD bottle probe for YSI® meters (5750)
Category: Dissolved Oxygen Probes and Sensors
Description: YSI
®
BOD probes
Brand:
YSI
Model Number:
5750
YO-05519-10
More Details
Self-stirring BOD bottle probe, 115 VAC, for YSI® meters (5905-115)
Category: Dissolved Oxygen Probes and Sensors
Description: YSI
®
BOD probes
Brand:
YSI
Model Number:
5905-115
YO-05519-42
More Details
YSI BOD probe adapter (5011)
Category: YSI Meter Accessories
Description: YSI DO Membrane Cap Probe
Brand:
YSI
Model Number:
5011
YO-05510-62
More Details
pH Probe for YSI Multiparameter Meters (5564)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
YSI
Model Number:
5564
YO-99852-50
More Details
pH/ORP Probe for YSI Multiparameter Meters (5565)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
YSI
Model Number:
5565
YO-99852-52
More Details
Replacement YSI DO membrane cap kit, 1.00 mil PTFE (5906)
Category: YSI Meter Accessories
Description: YSI DO Membrane Cap Probe
Brand:
PTFE
Model Number:
5906
YO-05519-61
More Details
YSI Oxygen Probe for YSI 5300A Monitors
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
YSI
Model Number:
5331A
YO-05520-16
More Details
Standard DO membrane kit for DO probe 05521-00, pack of 30 membranes a (5775)
Category: YSI Meter Accessories
Description: Replacement Membrane Kits
Brand:
YSI
Model Number:
5775
YO-05522-00
More Details
Replacement Self-Stirring BOD Probe for accumet Meters
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Accumet
YO-53109-50
More Details
Temperature Meter with Pipe Clamp for diameters from 1/4 to 3/8" and r (4005I)
Category: Handheld Thermocouple Meters
Description: Cooper/Atkins Cordless Temperature Meter
Brand:
Cooper-Atkins
Model Number:
4005I
YO-90025-03
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Field DO probe for YSI® meters (5739)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Field Case (waterproof) (YO-27444-82)
|
Next: Field tripod (840093)
31125329-p-content_2 31125329pcontent2