+GF+ Signet, Pv8S040, PVC Strap-On Saddle for 3" PVC 40/80 Pipe

YO-05620-19
Loading price...

Call for availability.
Qty
    Fittings are precision crafted to ensure proper sensor insertion depth and accurate flow measurement. Pipe fitting sizes range from 1/2" to 12" in diameter. Use our chemical compatibility database to select a fitting material compatible with your fluid. Other pipe sizes and materials are available on special order—please call our Application Specialists for more information.PVC Saddle Fittings for PVC 40 pipes from 2" to 4" in diameter; PVC 80 pipes from 6" to 8" in diameter. Specify schedule of pipe when ordering.
    METALEX, ROTOR-X—TM Signet Scientific
    Product Type Paddle Wheel Flowmeter
    Material PVC 40
    Model PV8S040





    Technical Resources
     
    Related Products for +GF+ Signet, Pv8S040, PVC Strap-On Saddle for 3" PVC 40/80 Pipe
    Image
      Product Detail
    SKU

    +GF+ Signet, Pv8S030, PVC Strap-On Saddle for 3" PVC 40/80 Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Saddle Fittings
    Brand: +GF+ Signet


    +GF+ Signet, Ir8S030, Iron Strap-on Saddle for 3" SCH 80 Metal Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: Galvanized Iron (Schedule 80) Saddles
    Brand: +GF+ Signet


    +GF+ Signet, PV8S020, PVC Strap-On Saddle for 2" PVC 40

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Saddle Fittings
    Brand: +GF+ Signet


    +GF+ Signet, Pv8S025, PVC Strap-On Saddle for 2.5" PVC 40/80 Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Saddle Fittings
    Brand: +GF+ Signet


    +GF+ Signet, Pv8S060, PVC Strap-On Saddle for 6" PVC 40/80 Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Saddle Fittings
    Brand: +GF+ Signet


    +GF+ Signet, Pv8S080, PVC Strap-On Saddle for 8" PVC 40/80 Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Saddle Fittings
    Brand: +GF+ Signet


    +GF+ Signet Conductivity/Resistivity Cell, 316SS, K = 10.0, 3/4" NPTM inline/submersion threads

    Category: Conductivity Electrodes - Industrial
    Description: +GF+ Signet Conductivity/Resistivity Electrodes
    Brand: +GF+ Signet


    +GF+ Signet, Cpv8T012F, CPVC Tee for 1-1/4" CPVC 80 Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Tee Fittings
    Brand: +GF+ Signet


    +GF+ Signet, Cpv8T015F, CPVC Tee for 1-1/2" CPVC 80 Pipe

    Category: Paddle Wheel Flowmeter Installation Fittings
    Description: PVC Tee Fittings
    Brand: +GF+ Signet


    GF Signet 2551 Insertion Magmeter, PP/SS, 0.5-4", No Display, S3L/Frequency

    Category: Magnetic Flow meters
    Description: +GF+ Signet Insertion Magmeter Flow Sensors
    Brand: +GF+ Signet


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    +GF+ Signet, Pv8S040, PVC Strap-On Saddle for 3" PVC 40/80 Pipe
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: +GF+ Signet, Pv8S030, PVC Strap-On Saddle for 3" PVC 40/80 Pipe  |  Next: +GF+ Signet, Pv8S060, PVC Strap-On Saddle for 6" PVC 40/80 Pipe