Ionic strength and pH adjustors, ISA 5 M NaNO3 (940011)
YO-05730-27
Loading price...
Usually ships in 14 days or sooner.
Qty
    Solution kits sold separately below include all of the solutions needed for accurate measurements. Each kit includes one reference filling pipette, one 60-mL bottle of reference fill solution, one 475-mL bottle of 1000 ppm standard solution, and one 475-mL bottle of ionic strength adjustor (except models 27502-91 and 27502-69, which come in a one-gallon bottle; model 27502-86 does not require an ionic strength adjustor).
    Replacement ionic strength adjustors come in 475-mL bottles (except model 27503-58, which comes in a one-gallon bottle).
    Product Type Ionic Strength/pH Adjusters
    Model 940011
    Manufacturer number 940011
    Technical Resources
    Related Products for Ionic strength and pH adjustors, ISA 5 M NaNO3 (940011)
    Image
      Product Detail
    SKU
    Ionic strength and pH adjustors, ISA 4 M KCl (932011)
    Category: Ion-Selective Electrodes Solutions
    Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
    Brand: Thermo Scientific Orion
    Ionic strength and pH adjustor; Interference suppressor (930710)
    Category: Ion-Selective Electrodes Solutions
    Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
    Brand: Thermo Scientific Orion
    Ionic strength and pH adjustor; SAOB reagent for S -2
    Category: Ion-Selective Electrodes Solutions
    Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
    Brand: Thermo Scientific Orion
    Ionic strength and pH adjustors, TISAB II (940909)
    Category: Ion-Selective Electrodes Solutions
    Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
    Brand: Thermo Scientific Orion
    Ionic strength and pH adjustors, TISAB III (conc.) (940911)
    Category: Ion-Selective Electrodes Solutions
    Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
    Brand: Thermo Scientific Orion
    Ionic strength and pH adjustor; Alkaline reagent (951011)
    Category: Ion-Selective Electrodes Solutions
    Description: Ionic Strength and pH Adjusters for Orion Ion-Selective Electrodes
    Brand: Thermo Scientific Orion
    Replacement ionic strength adjustors, 5 M NaCl (NH4IS01)
    Category: Ion-Selective Electrodes Solutions
    Description: Cole-Parmer Replacement ionic strength adjustors for Cole-Parmer Ion-Selective Electrodes
    Brand: Cole-Parmer
    Replacement ionic strength adjustors, 5 M NaNO 3
    Category: Ion-Selective Electrodes Solutions
    Description: Cole-Parmer Replacement ionic strength adjustors for Cole-Parmer Ion-Selective Electrodes
    Brand: Cole-Parmer
    Replacement ionic strength adjustors, 5 M NaCIO 4
    Category: Ion-Selective Electrodes Solutions
    Description: Cole-Parmer Replacement ionic strength adjustors for Cole-Parmer Ion-Selective Electrodes
    Brand: Cole-Parmer
    Replacement ionic strength adjustors, 4 M KCl (I4KCL00)
    Category: Ion-Selective Electrodes Solutions
    Description: Cole-Parmer Replacement ionic strength adjustors for Cole-Parmer Ion-Selective Electrodes
    Brand: Cole-Parmer
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    Ionic strength and pH adjustors, ISA 5 M NaNO3 (940011)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
    Previous: Ionic strength and pH adjustors, ISA 4 M KCl (932011)  |  Next: Ionic strength and pH adjustors, TISAB II (940909)