Electrode fill solution, 4 M KCl with Ag+. Case of five 60-mL bottles (900011)

YO-05730-42
Loading price...

Usually ships in 14 days or sooner.
Qty
    Thermo Scientific Orion filling solutions for use with pH electrodes and reference electrodes. Filling solutions are packaged as five 2 oz. (5 x 60mL) bottles.
    Product Type Accessory
    Brand Thermo Scientific Orion
    Model 900011
    Manufacturer number 900011





    Technical Resources
     
    Related Products for Electrode fill solution, 4 M KCl with Ag+. Case of five 60-mL bottles (900011)
    Image
      Product Detail
    SKU

    ROSS electrode fill solution, 3 M KCl. Case of five 60-mL bottles (810007)

    Category: Buffers and Solutions for pH/Ion Measurement
    Description: Thermo Scientific Orion Reference Fill Solutions
    Brand: Thermo Scientific Orion


    Oakton reference fill solution, 4 M KCl, two 125-mL bottles/pk

    Category: Buffers and Solutions for pH/Ion Measurement
    Description: Oakton Reference Fill Solutions
    Brand: Oakton


    Thermo Scientific Orion electrode fill solution, 3 M KCl, 5 x 60mL bottles

    Category: Buffers and Solutions for pH/Ion Measurement
    Description: Thermo Scientific Orion Reference Fill Solutions
    Brand: Thermo Scientific Orion


    Oakton reference fill solution, 3.8 M KCl with AgCl, two 125-mL bottles/pk

    Category: Buffers and Solutions for pH/Ion Measurement
    Description: Oakton Reference Fill Solutions
    Brand: Oakton


    Thermo Scientific- Orion ROSS Replacement Electrode Fill Solution, (4) x 4 oz. Bottles

    Category: pH/Ion Industrial Electrodes
    Description: Thermo Scientific- Orion 2001SC Ross Ultra Pure Water pH Electrode System
    Brand: Thermo Scientific Orion


    Thermo Scientific Orion pH electrode cleaning solution B, 4 x 60 mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion


    Thermo Scientific Orion pH electrode cleaning solution D, 4 x 60 mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion


    Reference fill solution, 4 M KCl saturated with AgCl, 125-mL bottle

    Category: pH/Ion Meter Accessories
    Description: Water Analysis Accessories
    Brand: Cole-Parmer


    Reference fill solution, 4 M KCl, 125-mL bottle

    Category: pH/Ion Meter Accessories
    Description: Water Analysis Accessories
    Brand: Cole-Parmer


    Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Storage Solutions
    Brand: Thermo Scientific Orion


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers

    Do you have questions about this product?

    get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      What is the shelf life of this product?
      Asked on 6/11/2012 by Anonymous

      1 answer

      • Staff Reviewer

        A:

        Shelf Life: 3-6 months after being opened. 2yrs if unopened.

        Answered on 6/11/2012 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews

    Electrode fill solution, 4 M KCl with Ag+. Case of five 60-mL bottles (900011)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Electrode Cleaning Polish, 25 Gram Bottle (YO-02656-40)  |  Next: Electrode Holder, 85 mm H x 13 mm diameter