Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Oakton high accuracy 7 000 pH buffer 500 mL case of 12
Click to enlarge
More Product Details
Specifications
Technical Resources
MSDS
Brochure
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-05942-47
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Buffers and Solutions for pH/Ion Measurement
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-05942-49
,
browse similar items
or
Contact us for more information.
Oakton® high-accuracy 7.000 pH buffer, 500-mL, case of 12
YO-05942-47
Loading price...
Discontinued
Qty
:
Save to shopping list
Buffers for meters with 0.001 resolution are accurate to either ±0.002 pH unit (pH 4 and 7 buffers) or ±0.005 pH unit (pH 10 buffer) when referenced to NIST-traceable reference materials at 77°F (25°C). Available as an individual 500-mL bottle, or in a case of 12.
Specifications
Product Type
pH Buffers
Description
pH 7.000
Volume (mL)
500
Quantity
500 mL, case of 12
Qty/pk
12
Brand
Oakton
Manufacturer number
R0594246-12CS
Technical Resources
MSDS 1 (135 B)
Oakton Catalogo en Espanol.pdf (8.3 MB)
Related technical articles (Roll over for summary)
Tips for pH Buffer Use
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Certificate of Analysis
Enter lot number
Sample C of A
Related Searches for Oakton® high-accuracy 7.000 pH buffer, 500-mL, case of 12
Oakton High Accuracy Ph Buffer Solution
Oakton High Accuracy Rtd Probes
Ph Buffer Packets 12 13
High Accuracy Dissolved Ozone Meters
Oakton 4.01 Ph Buffer
7.00 Ph Buffer
500 Ml Volumetric Flask
500 Ml Round Bottom Flask
Oakton Ph Buffer Solution Msds
Oakton Ph Buffer Coa
Related Products for Oakton® high-accuracy 7.000 pH buffer, 500-mL, case of 12
Image
Product Detail
SKU
Oakton® high-accuracy 4.001 pH buffer, 500-mL, case of 12
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton® High Accuracy pH Buffer Solution
Brand:
Oakton
YO-05942-27
More Details
Oakton® high-accuracy 10.000 pH buffer, 500-mL, case of 12
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton® High Accuracy pH Buffer Solution
Brand:
Oakton
YO-05942-67
More Details
Oakton 4.01 pH buffer, 500-mL, case of 12 (YO-05942-21)
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-05942-21
More Details
Oakton 7.00 pH buffer, 500-mL, case of 12 (YO-05942-41)
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-05942-41
More Details
Oakton 10.00 pH buffer, 500-mL, case of 12 (YO-05942-61)
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-05942-61
More Details
Oakton 4.01 pH buffer, 500-mL
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-00654-00
More Details
Oakton 1.68 pH buffer, 500-mL
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-00654-01
More Details
Oakton 6.86 pH buffer, 500-mL
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-00654-03
More Details
Oakton 7.00 pH buffer, 500-mL
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-00654-04
More Details
Oakton 9.18 pH buffer, 500 mL
Category: Buffers and Solutions for pH/Ion Measurement
Description: Oakton pH Buffer Solutions
Brand:
Oakton
YO-00654-07
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Oakton® high-accuracy 7.000 pH buffer, 500-mL, case of 12
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Oakton® high-accuracy 4.001 pH buffer, 500-mL, case of 12
|
Next: Oakton® Infrared Thermometer with Type-K Input (High Temp) (YO-39644-10)
31125329-p-content_2 31125329pcontent2