Home  >  Mechanical Air Velocity Meters >
  

TSI/Alnor Velometer Jr Intrinsically Safe Air Velocity Meter; 800 ft/min

YO-05949-60
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Mechanical design allows measurements without a power source
  • Intrinsically safe meter allows safe use in hazardous environments
Compact meters can be used almost anywhere—fume hoods, spray booths, drying tunnels, HVAC systems. Easy to operate—simply hold the meter in air stream and read the scale. Turn the orifice fitting to switch ranges. Choose from six dual-range units that read in either feet/minute or meters/second.
What's Included:
certificate of NIST traceability and carrying case.
Product Type Vane Anemometer
Range 0 to 200, 0 to 800 ft/min
Accuracy ±5% of full scale
Display analog
Dimensions 3 3/4" W x 3 1/8" H x 1 1/8" D
Brand Alnor
Model 8100-8
Manufacturer number 8100-8

Calibrations
Qty/Price
Availability

EACH
Usually ships in 25 days.




Technical Resources
 
Related Products for TSI/Alnor Velometer Jr Intrinsically Safe Air Velocity Meter; 800 ft/min
Image
  Product Detail
SKU

TSI ™ VelociCalc Air Velocity Meter, Remote Probe

Category: Hot-Wire Anemometers
Description: TSI VelociCalc Air Velocity Meters
Brand: TSI


Alnor mechanical air velocity meter, 0-1600 ft/min

Category: Mechanical Air Velocity Meters
Description: Alnor Air Velocity Meters
Brand: Alnor


TSI ™ VelociCalc Air Velocity Meter with Straight Probe, no Humidity Sensor

Category: Hot-Wire Anemometers
Description: TSI VelociCalc Air Velocity Meters
Brand: TSI


TSI ™ VelociCalc Air Velocity Meter with Straight Probe and Humidity Sensor

Category: Hot-Wire Anemometers
Description: TSI VelociCalc Air Velocity Meters
Brand: TSI


TSI 9535-A VelociCalc Air Velocity Meter; Articulating Probe/Manual Log

Category: Hot-Wire Anemometers
Description: TSI VelociCalc Air Velocity Meters
Brand: TSI


TSI 9545-A VelociCalc Air Velocity Meter; Articulating Probe/Auto Log

Category: Hot-Wire Anemometers
Description: TSI VelociCalc Air Velocity Meters
Brand: TSI


TSI/Alnor Velometer Jr Intrinsically Safe Air Velocity Meter; 2500 ft/min

Category: Mechanical Air Velocity Meters
Description: Alnor Air Velocity Meters
Brand: Alnor


TSI Alnor AVM410 Thermoanemometer Meter Kit

Category: Hot-Wire Anemometers
Description: TSI Alnor Basic Thermoanemometer Meter
Brand: Alnor


TSI Alnor HM675 Hydronic Manometer Water Air Pressure Meter

Category: Digital and Analog Manometers
Description: Alnor Hydronic Manometers
Brand: Alnor


TSI Alnor HM685 Hydronic Manometer Water Air Flow Meter

Category: Digital and Analog Manometers
Description: Alnor Hydronic Manometers
Brand: Alnor


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

TSI/Alnor Velometer Jr Intrinsically Safe Air Velocity Meter; 800 ft/min
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: TSI/Alnor Velometer Jr Intrinsically Safe Air Velocity Meter; 2500 ft/min  |  Next: TT-103R-W : Soil & Water Temp Sensor Analog 4-20mA