Subtotal =  
Items =  
View Cart
Welcome
 
Home  >  Mechanical Air Velocity Meters   

Velometer Jr. 0/200 and 0/800 FPM Includes Carrying Case, NIST Certifi (8100-8)

YO-05949-60
Loading price...

Call for availability.
Qty

  • Mechanical design allows measurements without a power source
  • Intrinsically safe meter allows safe use in hazardous environments
Compact meters can be used almost anywhere—fume hoods, spray booths, drying tunnels, HVAC systems. Easy to operate—simply hold the meter in air stream and read the scale. Turn the orifice fitting to switch ranges. Choose from six dual-range units that read in either feet/minute or meters/second.
What's Included:
certificate of NIST traceability and carrying case.
Product Type Vane Anemometer
Range 0 to 200, 0 to 800 ft/min
Accuracy ±5% of full scale
Display analog
Dimensions 3 3/4" W x 3 1/8" H x 1 1/8" D
Manufacturer number 8100-8
Brand Alnor
Model 8100-8

Calibrations
Qty/Price
Availability

EACH
Usually ships in 26 days.
Reconditioned & Like New
Qty/Price
Availability
RECOND 05949-60
EW-00315DI

at Call for Price

Usually ships in 14 days.




Technical Resources
 
Related Products for Velometer Jr. 0/200 and 0/800 FPM Includes Carrying Case, NIST Certifi (8100-8)
Image
  Product Detail
SKU

Velometer Jr. 0/500 and 0/2500 FPM Includes Carrying Case, NIST Certif (8100B-25)

Category: Mechanical Air Velocity Meters
Description: Alnor Air Velocity Meters
Brand: Alnor
Model Number: 8100B-25


Carrying case for Digi-Sense meter (1316-1)

Category: Thermocouple Accessories
Description: Carrying Cases - for Handheld Meters
Brand: Digi-Sense
Model Number: 1316-1


Cordura carrying case (PCCP0007 - 100 PC REL.)

Category: Thermocouple Accessories
Description: Carrying Cases - for Handheld Meters
Brand: Cordura
Model Number: PCCP0007 - 100 PC REL.


Fluke® CXT80 Hard Carrying Case

Category: Fluke® 87V IMSK Industrial Service Combo Multimeter Kit
Description: Fluke® Hard Carrying Case for 80V-Series Industrial Troubleshooting Multimeters
Brand: Fluke
Model Number: CXT80


Fluke Calibration 7526A-CASE Carrying Case for the 7526A Precision Pro

Category: Calibration Equipment
Description: Fluke Calibration 7526A-CASE Carrying Case for the 7526A Precision Process Calibrator
Brand: Fluke


IO800A : Soft fabric carrying case with accessory pocket (DPI 800)

Category: Calibrators
Description: Calibrators Accessories
Brand: Druck
Model Number: DPI 800


Carrying case for Acorn meters. Includes one rinse bottle and three so (YO-35632-97)

Category: Accessories for pH/Ion Meters
Description: Water Analysis Accessories
Brand: Acorn


5170 Gaussmeter W/4in Probe Ac Adapter Hard Carrying Case (5170)

Category: Gauss Meters
Description: Pacific Scientific OECO Gauss - Tesla Meters
Brand: FW Bell
Model Number: 5170


YSI Soft-Sided Carrying Case for Professional Series Meters

Category: Accessories for pH/Ion Meters
Description: Water Analysis Accessories
Brand: Lab Products
Model Number: 603075


YSI Hard-Sided Carrying Case for Professional Series Meters

Category: Accessories for pH/Ion Meters
Description: Water Analysis Accessories
Brand: Lab Products
Model Number: 603074


Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews