Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Polypropylene tri corner beaker assortment pack
Click to enlarge
More Product Details
Specifications
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-06010-05
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Plastic Beakers
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-06010-07
,
browse similar items
or
Contact us for more information.
Polypropylene tri-corner beaker assortment pack (80091)
YO-06010-05
Loading price...
Discontinued
Qty
:
Save to shopping list
Polypropylene beakers feature a tri-corner design that provides three dripless pouring spouts. Beakers are perfect for use with sturrers- flat bottom keeps stir bars centered. Graduated with an accuracy or ±5%. Withstands temperatures to 250°F (121°C). Autoclavable.
Specifications
Product Type
Beaker
Autoclavable
Yes
Withstands Temp (high) (°F)
275
Withstands Temp (low) (°F)
32
Quantity
30
Material
Polypropylene
Technical Resources
Related Searches for Polypropylene tri-corner beaker assortment pack (80091)
Cole-Parmer Tri-Corner Beakers
Tri Cornered Polypropylene Beakers
Cole-Parmer Heat Shrink Assortment Packs
Cole-Parmer Fittings Assortment Packs and Kits
Fittings Assortment Packs
Fittings Assortment Packs and Kits
Glass Beakers 600ml Pack of 8
Tri Pour Polypropylene Beakers
Plasticware Beakers
Small Plastic Lab Beakers
Related Products for Polypropylene tri-corner beaker assortment pack (80091)
Image
Product Detail
SKU
Polypropylene tri-corner beaker 30/pack
Category: Plastic Beakers
Description: Cole-Parmer TRI-CORNER BEAKERS,PP
Brand:
Cole-Parmer
Model Number:
BTCST30
YO-06010-07
More Details
Polypropylene tri-corner beaker, 50 mL (80092)
Category: Plastic Beakers
Description: Cole-Parmer TRI-CORNER BEAKERS,PP
Brand:
Cole-Parmer
Model Number:
80092
YO-06010-35
More Details
Cole-Parmer Tri-Corner Beaker, PP, 50mL, 100/pk
Category: Plastic Beakers
Description: Cole-Parmer Tri-Corner Beakers, PP
Brand:
Cole-Parmer
YO-06020-01
More Details
Cole-Parmer Tri-Corner Beaker, PP, 100mL, 100/pk
Category: Plastic Beakers
Description: Cole-Parmer Tri-Corner Beakers, PP
Brand:
Cole-Parmer
YO-06020-03
More Details
Cole-Parmer Tri-Corner Beaker, PP, 250mL, 100/pk
Category: Plastic Beakers
Description: Cole-Parmer Tri-Corner Beakers, PP
Brand:
Cole-Parmer
YO-06020-05
More Details
Cole-Parmer Tri-Corner Beaker, PP, 400mL, 100/pk
Category: Plastic Beakers
Description: Cole-Parmer Tri-Corner Beakers, PP
Brand:
Cole-Parmer
YO-06020-07
More Details
Cole-Parmer Tri-Corner Beaker, PP, 800mL, 100/pk
Category: Plastic Beakers
Description: Cole-Parmer Tri-Corner Beakers, PP
Brand:
Cole-Parmer
YO-06020-09
More Details
Cole-Parmer Tri-Corner Beaker, PP, 1000mL, 100/pk
Category: Plastic Beakers
Description: Cole-Parmer Tri-Corner Beakers, PP
Brand:
Cole-Parmer
YO-06020-11
More Details
Azlon polypropylene "square ratio" beaker assortment pack; one each of
Category: Plastic Beakers
Description: Azlon Tapered Beakers, PP
Brand:
Azlon
Model Number:
522125
YO-66201-90
More Details
T-connector assortment pack, PP, 6 each of sizes 4mm to 14 mm, 36 piec (YO-06294-82)
Category: Fittings Assortment Packs and Kits
Description: Fittings Assortment Packs
Brand:
Cole-Parmer
YO-06294-82
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Polypropylene tri-corner beaker assortment pack (80091)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Polypropylene tri-corner beaker 30/pack
|
Next: Polypropylene tri-corner beaker, 50 mL (80092)
31125329-p-content_2 31125329pcontent2