Wide Mouth Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (3303)

YO-06148-14
Loading price...

Call for availability.
Qty

  • Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes—each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.
Product Type Mailers
Material Fiberboard
Capacity (Liters) 0.03
Capacity (mL) 30
Model 3303
Brand Qorpak
Manufacturer number PLC-03558





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for Wide Mouth Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (3303)
    Image
      Product Detail
    SKU

    Clear wide mouth glass jar, 1 oz, 48 per case (15714304-48N)

    Category: Bottles
    Description: Clear Wide Mouth Bottles, 1oz
    Brand: Cole-Parmer


    Wide-Mouth Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs (3304)

    Category: Fiberboard Mailers
    Description: Fiberboard Mailer with wide-mouth HDPE bottle
    Brand: Qorpak


    Wide-Mouth Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs (3305)

    Category: Fiberboard Mailers
    Description: Fiberboard Mailer with wide-mouth HDPE bottle
    Brand: Qorpak


    Thermo Scientific Nalgene fluorinated wide-mouth bottle, 1/2 gallon

    Category: Wide-Mouth Bottles
    Description: Thermo Scientific Nalgene Fluorinated 1/2-Gallon Wide-Mouth Bottle
    Brand: Thermo Scientific Nalgene


    Boston Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (2300)

    Category: Fiberboard Mailers
    Description: Fiberboard Mailer with Boston round glass bottle
    Brand: Qorpak


    Medium Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (2304)

    Category: Fiberboard Mailers
    Description: Fiberboard Mailer with medium round glass bottle
    Brand: Qorpak


    Clear square wide mouth bottle, 1 oz, 48 per case (15712404-48N)

    Category: Bottles
    Description: Clear Wide Mouth Bottles, 1oz
    Brand: Cole-Parmer


    Thermo Scientific Nalgene polypropylene wide-mouth bottle, 4 L

    Category: Wide-Mouth Plastic Bottles
    Description: Thermo Scientific Nalgene Polypropylene Wide-Mouth Bottles
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene fluorinated wide-mouth bottle, 1 L

    Category: Wide-Mouth Bottles
    Description: Thermo Scientific Nalgene Fluorinated Wide-Mouth Bottles
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene amber high-density polyethylene wide-mouth bottle, 1 L

    Category: Wide-Mouth Bottles
    Description: Thermo Scientific Nalgene Amber Wide-Mouth Bottle, HDPE
    Brand: Thermo Scientific Nalgene


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Wide Mouth Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs (3303)
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Whole Range pH Test Paper; pH Range 1 to 14; 9 mm x 5.5 m Roll  |  Next: Wide-Mouth Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs (3304)