Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Tear away lid for 06274 00 1 gallon
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-06274-32
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Tear away lid for 06274-00 - 1 gallon (C1W/T)
YO-06274-32
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for Tear away lid for 06274-00 - 1 gallon (C1W/T)
1 Gallon Bottles
1 Gallon Container
1 Gallon Bottle Pump Agar
Sodium Silicate Solution 1 Gallon
1 Gallon Fluorinated Hdpe Bottles
1 Gallon Thermos
Glas Col 1 Gallon Heating Pot
1 Gallon Float Switch
1 Gallon Hdpe Containers
1 Gallon Air Tank
Related Products for Tear away lid for 06274-00 - 1 gallon (C1W/T)
Image
Product Detail
SKU
Tear away lid with spout for 06274-15, 3-12 gal or -25 five gal pail (C5W/PS)
Category: Gallon HDPE FDA and USDA Pails
Description: Tear away lid with spout
Brand:
Cole-Parmer
Model Number:
C5W/PS
YO-06274-42
More Details
Tear away lid for 06274-03 - 2 gallon (C2W/T)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
C2W/T
YO-06274-34
More Details
Tear away lid for 06274-15, -25 or -27 (C5W/T)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
C5W/T
YO-06274-36
More Details
Hinged-Lid Sample Containers, PP, 1/4 oz, 2500/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-00
More Details
Bain Marie Container Snap-Tight, Positive-Lock Lid, HDPE, 63530-00, -0
Category: Plastic Containers
Description: Bain Marie Container Lids, HDPE
Brand:
Lab Products
Model Number:
H3241200
YO-63530-50
More Details
Snap-On Lid, PP, Translucent, for 1-L containers, 12/pk
Category: Plastic Containers
Description: Snap-On Lids for Round Containers with Handles, PP
Brand:
Lab Products
Model Number:
1077030
YO-63571-20
More Details
Hinged-Lid Sample Containers, PP, 1/2 oz, 2000/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-10
More Details
Hinged-Lid Sample Containers, PP, 1 oz, 1300/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-30
More Details
Stackable high-density polyethylene pail, 1 gallon (1W 0627400)
Category: Buckets and Pails
Description: Gallon HDPE FDA and USDA Pails
Brand:
Cole-Parmer
Model Number:
1W 0627400
YO-06274-00
More Details
Replacement Classifier Lid (500 051 00)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
500 051 00
YO-02658-72
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Tear away lid for 06274-00 - 1 gallon (C1W/T)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: TDS5104B: 4-Channel, 1 GHz, up to 5 GS/s, 16M Record Length (TDS5104B)
|
Next: Tear away lid for 06274-03 - 2 gallon (C2W/T)
31125329-p-content_2 31125329pcontent2