Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-06275-03
Square high-density polyethylene lid for 06275-03
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-06275-03
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Buckets and Pails
Share
|
Square high-density polyethylene lid for 06275-03 (C4WS/T)
YO-06275-03
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Square design allows better use of storage space
Manufactured from 100% prime virgin high density polyethylene (HDPE), these pails are manufactured in one piece and feature a heavy duty plastic easy grip handle.
The lid, sold separately (#06275-03), features a tamper resistant ''tear-tab'' that allows easy access to product. No lid opener is required. FDA approved.
Specifications
Product Type
Bucket
Quantity
1
Material
HDPE
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related Searches for Square high-density polyethylene lid for 06275-03 (C4WS/T)
Nalgene High-Density Polyethylene (Hdpe) Beakers
Cole-Parmer High-Density Polyethylene Utility Funnels
Chemical High Density Polyethylene Hdpe Bottles
10 L High Density Polyethylene Carboys
High Density Polyethylene Carboy Rect
Rectangular Bottles High Density Polyethylene
Un-Approved High-Density Polyethylene Carboy with Handle
High Density Polyethylene Bottles
Thermo Scientific High-Density Polyethylene Jars
High Density Polyethylene Tubings
Related Products for Square high-density polyethylene lid for 06275-03 (C4WS/T)
Image
Product Detail
SKU
UN-approved square high-density polyethylene carboy with handle,5 gall (16314500-8) (YO-06065-31)
Category: Discontinued Items
Description: UN-Approved High-Density Polyethylene Carboy with Handle
Brand:
Cole-Parmer
Model Number:
16314500-8
YO-06065-31
More Details
Large industrial-size high-density polyethylene funnel, 18 L (YO-06121-36)
Category: Industrial Funnels
Description: High-density polyethylene (HDPE) Funnels
Brand:
Guaranteed In Stock Program
YO-06121-36
More Details
High-density polyethylene utility funnel, 16 oz (YO-06122-40)
Category: Utility Funnels
Description: High-density polyethylene Utility Funnels
Brand:
Cole-Parmer
YO-06122-40
More Details
High-density polyethylene utility funnel, 32 oz (YO-06122-50)
Category: Utility Funnels
Description: High-density polyethylene Utility Funnels
Brand:
Cole-Parmer
YO-06122-50
More Details
High-density polyethylene utility funnel, 64 oz (YO-06122-60)
Category: Utility Funnels
Description: High-density polyethylene Utility Funnels
Brand:
Cole-Parmer
YO-06122-60
More Details
High-Density Polyethylene (HDPE) Tubing, 1/8 x 1/4", 100 ft/pack
Category: Rigid Tubing
Description: High-Density Polyethylene (HDPE) Tubing
Brand:
Guaranteed In Stock Program
YO-95501-00
More Details
High-Density Polyethylene (HDPE) Tubing, 0.17 x 1/4", 100 ft/pack
Category: Rigid Tubing
Description: High-Density Polyethylene (HDPE) Tubing
Brand:
Cole-Parmer
YO-95501-02
More Details
High-Density Polyethylene (HDPE) Tubing, 3/16 x 5/16", 100 ft/pack
Category: Rigid Tubing
Description: High-Density Polyethylene (HDPE) Tubing
Brand:
Cole-Parmer
YO-95501-04
More Details
High-Density Polyethylene (HDPE) Tubing, 1/4 x 3/8", 100 ft/pack
Category: Rigid Tubing
Description: High-Density Polyethylene (HDPE) Tubing
Brand:
Cole-Parmer
YO-95501-06
More Details
High-Density Polyethylene (HDPE) Tubing, 5/16 x 7/16", 100 ft/pack
Category: Rigid Tubing
Description: High-Density Polyethylene (HDPE) Tubing
Brand:
Cole-Parmer
YO-95501-08
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Square high-density polyethylene lid for 06275-03 (C4WS/T)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Square high-density polyethylene jug, 2 L (16706419-50N)
|
Next: Square high-density polyethylene pail, 4-1/4 gallon (4WS)
31125329-p-content_2 31125329pcontent2