Thermo Scientific Nalgene, 1/4" Grid Case Bound Paper Lab Notebook; 1/Pk

YO-06553-00
Loading price...

Call for availability.
Qty
    Laboratory notebooks feature numbered pages printed with either a green grid or green horizontal lines; additional pages include issuance page, instructions for keeping research records, and table of contents. Numbered pages have spaces for signatures and dates. Choose either Polypaper® or conventional paper. Uncoated Polypaper® is highly resistant to moisture, chemicals, tearing, and aging; works best with ballpoint pen or felt-tip marker.
    Product Type Notebooks
    Description 8-1/2" x 11" with 1/4" grid, green
    No. of pages 100
    Brand Thermo Scientific Nalgene
    Model 6301-1000
    Manufacturer number 6301-1000





    Technical Resources
     
    Related Products for Thermo Scientific Nalgene, 1/4" Grid Case Bound Paper Lab Notebook; 1/Pk
    Image
      Product Detail
    SKU

    Thermo Scientific Nalgene, 1/4" Grid Polypaper Lab Notebook; 1/Pk

    Category: Notebooks
    Description: Lab Notebooks
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Filter Units, Gridded 0.45um CN, 115mL; 12/Pk

    Category: Thermo Scientific Nalgene Disposable Filterware
    Description: Thermo Scientific Nalgene Cellulose Nitrate (CN) Filter Units
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Filter Units, Gridded 0.45um CN, 500mL; 12/Pk

    Category: Thermo Scientific Nalgene Disposable Filterware
    Description: Thermo Scientific Nalgene Cellulose Nitrate (CN) Filter Units
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Filter Units, Gridded 0.2um CN, 1000mL; 12/Pk

    Category: Thermo Scientific Nalgene Disposable Filterware
    Description: Thermo Scientific Nalgene Cellulose Nitrate (CN) Filter Units
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Filter Units, Gridded 0.2um PES, 115mL; 72/Pk

    Category: Thermo Scientific Nalgene Disposable Filterware
    Description: Thermo Scientific Nalgene Polyethersulfone (PES) Filter Units and Bottletop Filters
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles, 8 mL, 12/Pk

    Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
    Description: Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles, 15 mL, 12/Pk

    Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
    Description: Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles, 30 mL, 12/Pk

    Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
    Description: Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles, 60 mL, 12/Pk

    Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
    Description: Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles
    Brand: Thermo Scientific Nalgene


    Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles, 250 mL, 12/Pk

    Category: Thermo Scientific Nalgene Narrow-Mouth Bottles
    Description: Thermo Scientific Nalgene Amber Narrow-Mouth HDPE Bottles
    Brand: Thermo Scientific Nalgene


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers

    Do you have questions about this product?

    get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      What are the covers made of? Are they resistant as well?
      Asked on 6/14/2011 by Joe from Richmond, VA

      1 answer

      • Staff Reviewer

        A:

        The cover is made of polyethylene, which are water proof.

        Answered on 6/16/2011 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought

    Thermo Scientific Nalgene, 1/4" Grid Case Bound Paper Lab Notebook; 1/Pk
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Thermo Scientific Nalgene Wraparound Face Shield, Polycarbonate  |  Next: Thermo Scientific Nalgene, 1/4" Grid Polypaper Lab Notebook; 1/Pk