Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Thermo Scientific Nalgene straight sided polypropylene wide mouth jar 60 mL
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-06758-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Jars
Share
|
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0002)
YO-06758-52
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Jars
Cap material
Polypropylene
Cap/stopper size
53 mm
Material
Polypropylene
Capacity (mL)
60
Qty/pk
12
Autoclavable
Yes
Dimensions
1-7/8" H x 1-7/8" OD
(4.8 x 4.8 cm)
Model
2118-0002
Brand
Thermo Scientific Nalgene
Manufacturer number
2118-0002
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related Searches for Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0002)
Thermo Scientific Nalgene Heavy-Duty Polypropylene Bottles
Thermo Scientific Nalgene Wide-Mouth Bottles
Thermo Scientific Nalgene Narrow-Mouth Style
Thermo Scientific Nalgene Hdpe Sample Bottles
Thermo Scientific Nalgene Polypropylene Jars
Thermo Scientific Nalgene Plastic Volumetric Flasks
Thermo Scientific Nalgene High-Density Polyethylene Bottles
Thermo Scientific Nalgene Amber Polypropylene Bottles
Thermo Scientific Nalgene 689 Polypropylene Tubing
Thermo Scientific Nalgene Polypropylene Trays
Related Products for Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0002)
Image
Product Detail
SKU
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-9050)
Category: Jars
Description: Polypropylene jars
Brand:
Thermo Scientific Nalgene
Model Number:
2118-9050
YO-06758-48
More Details
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0001)
Category: Jars
Description: Polypropylene jars
Brand:
Thermo Scientific Nalgene
Model Number:
2118-0001
YO-06758-50
More Details
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0004)
Category: Jars
Description: Polypropylene jars
Brand:
Thermo Scientific Nalgene
Model Number:
2118-0004
YO-06758-54
More Details
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0008)
Category: Jars
Description: Polypropylene jars
Brand:
Thermo Scientific Nalgene
Model Number:
2118-0008
YO-06758-56
More Details
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0016)
Category: Jars
Description: Polypropylene jars
Brand:
Thermo Scientific Nalgene
Model Number:
2118-0016
YO-06758-58
More Details
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0032)
Category: Jars
Description: Polypropylene jars
Brand:
Thermo Scientific Nalgene
Model Number:
2118-0032
YO-06758-60
More Details
Thermo Scientific Nalgene low-density polyethylene wide-mouth bottle, (2103-0002)
Category: Wide-Mouth Bottles to 1000 mL
Description: Thermo Scientific Nalgene Wide-Mouth Bottles, LDPE
Brand:
Thermo Scientific Nalgene
Model Number:
2103-0002
YO-06043-20
More Details
Thermo Scientific Nalgene high-density polyethylene wide-mouth bottle, (2104-0002)
Category: Sample Bottles
Description: Thermo Scientific Nalgene Wide-Mouth High-Density Polyethylene Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2104-0002
YO-06047-20
More Details
Thermo Scientific Nalgene high-density polyethylene wide-mouth jar, 2
Category: Wide-Mouth Plastic Bottles
Description: Thermo Scientific Nalgene High-density polyethylene bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2120-0005
YO-06055-10
More Details
Thermo Scientific Nalgene high-density polyethylene wide-mouth jar, 4
Category: Wide-Mouth Plastic Bottles
Description: Thermo Scientific Nalgene High-density polyethylene bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2120-0010
YO-06055-20
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0002)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0001)
|
Next: Thermo Scientific Nalgene straight-sided polypropylene wide-mouth jar, (2118-0004)
31125329-p-content_2 31125329pcontent2