Keck Standard Taper Clips, joint size 19, Blue, Pack of 12

YO-06840-10
Loading price...

Usually ships in 14 days or sooner.
Qty

  • Our most economical standard taper clips
These economical KECK adapters, clamps, and clips will not scratch glass or corrode—ideal for glass and tubing assembly. All KECK products are color-coded for fast, easy size identification. Use quick disconnect adapters for safe, convenient tubing-to-tubing or tubing-to-glass connections. Use RAMP CLAMP tubing clamps for easy, one-handed tubing flow control. Select standard taper clips (in plastic or lacquer-coated steel) or ball-joint clips for secure glass-to glass connections.These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°F (150°C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
Hot Tips:
The joint size on glassware is a two-part number, such as 24/40. To select the correct KECK ®  connector, match the first number on the glassware to the KECK joint size number.
Product Type Keck Clips
Color Blue
Manufacturer number MAKE-UP
Model MAKE-UP
Brand Duran





Technical Resources
 
Related Products for Keck Standard Taper Clips, joint size 19, Blue, Pack of 12
Image
  Product Detail
SKU

Keck Standard Taper Clips, joint size 10, green, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 14, Yellow, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 24, green, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 29, red, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 34, Orange, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 40, Yellow, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 45, Brown, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips, joint size 12, Violet, Pack of 12

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clips Assortment, 24/pack

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Keck Standard Taper Clip Assortment, Pack of 27

Category: Keck Clamps and Clips
Description: Keck Plastic Clips
Brand: Duran
Model Number: MAKE-UP


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought